Skip to Content
Merck
All Photos(1)

Documents

AV48884

Sigma-Aldrich

Anti-NSUN3 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-FLJ22109, Anti-FLJ22609, Anti-MST077, Anti-MSTP077, Anti-NOL1/NOP2/Sun domain family, member 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

38 kDa

species reactivity

rat, bovine, pig, rabbit, human, dog, mouse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NSUN3(63899)

General description

Mutations in NOP2/Sun domain family, member 3 (NSUN3) gene have been reported to be associated with lung cancer in non-smokers.

Immunogen

Synthetic peptide directed towards the C terminal region of human NSUN3

Application

Anti-NSUN3 antibody produced in rabbit is suitable for western blotting at a concentration of 0.5μg/ml.

Biochem/physiol Actions

NSUN3 belongs to the methyltransferase superfamily. RsmB/NOP family. It may have S-adenosyl-L-methionine-dependent methyl-transferase activity.

Sequence

Synthetic peptide located within the following region: LPLLQIELLRSAIKALRPGGILVYSTCTLSKAENQDVISEILNSHGNIMP

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bastien Job et al.
PloS one, 5(12), e15145-e15145 (2010-12-15)
Lung cancer in never smokers would rank as the seventh most common cause of cancer death worldwide. We performed high-resolution array comparative genomic hybridization analysis of lung adenocarcinoma in sixty never smokers and identified fourteen new minimal common regions (MCR)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service