Skip to Content
Merck
All Photos(2)

Key Documents

AV46165

Sigma-Aldrich

Anti-STIP1 (AB2) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-HOP, Anti-IEF-SSP-3521, Anti-P60, Anti-STI1, Anti-STI1L, Anti-Stress-induced-phosphoprotein 1 (Hsp70/Hsp90-organizing protein)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$2,017.00

R$2,017.00


Estimated to ship onMay 28, 2025



Select a Size

Change View
100 μL
R$2,017.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$2,017.00


Estimated to ship onMay 28, 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

63 kDa

species reactivity

bovine, rat, horse, mouse, rabbit, human, dog

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... STIP1(10963)

General description

Stress-induced-phosphoprotein 1 (Hsp70/Hsp90-organizing protein) (STIP1, HOP, IEF-SSP-3521, STI1, STI1L, P60) is a co-chaperone that regulates and assists heat shock proteins (major chaperones). STIP1 links the chaperones Hsp70 and Hsp90 together to facilitate their function. HOP ensures the productive folding of substrate proteins by controlling the chaperone activities of HSP70 and HSP90.

Specificity

Anti-STIP1 (AB2) polyclonal antibody reacts with bovine, human, mouse, rat, and canine stress-induced-phosphoprotein 1 (Hsp70/Hsp90-organizing protein) proteins..

Immunogen

Synthetic peptide directed towards the C terminal region of human STIP1

Application

Anti-STIP1 (AB2) polyclonal antibody is used to tag stress-induced-phosphoprotein 1 (Hsp70/Hsp90-organizing protein) for detection and quantitation by Western blotting and in plasma by immunohistochemical (IHC) techniques. It is used as a probe to determine the roles of stress-induced-phosphoprotein 1 (Hsp70/Hsp90-organizing protein) in the management of protein refolding by heat shock proteins such as Hsp70 and Hsp90.

Biochem/physiol Actions

STIP1 mediates the association of the molecular chaperones HSC70 and HSP90 (HSPCA and HSPCB).

Sequence

Synthetic peptide located within the following region: YQKALDLDSSCKEAADGYQRCMMAQYNRHDSPEDVKRRAMADPEVQQIMS

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service