Skip to Content
Merck
All Photos(1)

Key Documents

AV46038

Sigma-Aldrich

Anti-MTHFD2 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 2, Anti-NMDMC

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$2,017.00

R$2,017.00


Estimated to ship onMay 29, 2025



Select a Size

Change View
100 μL
R$2,017.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$2,017.00


Estimated to ship onMay 29, 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

35 kDa

species reactivity

human, rabbit, dog, bovine, mouse, rat, horse, guinea pig

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MTHFD2(10797)

Immunogen

Synthetic peptide directed towards the N terminal region of human MTHFD2

Application

Anti-MTHFD2 antibody produced in rabbit is suitable for western blotting at a concentration of 1.25μg/ml.

Biochem/physiol Actions

MTHFD2 encodes a mitochondrial bifunctional enzyme methylenetetrahydrofolate dehydrogenase/cyclohydrolase with NAD-dependent methylenetetrahydrofolate dehydrogenase and methenyltetrahydrofolate cyclohydrolase domain. Decrease in MTHFD2 expression leads to impairment of cell migration and invasion into extracellular matrix and reduces the cell fraction with a high CD44 expression. It facilitates the regulation of breast cancer cell migration and invasion.

Sequence

Synthetic peptide located within the following region: LPLPEHIDERRICNAVSPDKDVDGFHVINVGRMCLDQYSMLPATPWGVWE

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laura Lehtinen et al.
Oncotarget, 4(1), 48-63 (2013-01-09)
Vimentin is an intermediate filament protein, with a key role in the epithelial to mesenchymal transition as well as cell invasion, and it is often upregulated during cancer progression. However, relatively little is known about its regulation in cancer cells.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service