Skip to Content
Merck
All Photos(1)

Key Documents

AV44904

Sigma-Aldrich

Anti-FAM3C antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Family with sequence similarity 3, member C, Anti-GS3786, Anti-ILEI

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$3,345.00

R$3,345.00


Check Cart for Availability


Select a Size

Change View
100 μL
R$3,345.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$3,345.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

22 kDa

species reactivity

dog, rat, human, guinea pig, bovine

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FAM3C(10447)

Immunogen

Synthetic peptide directed towards the C terminal region of human FAM3C

Application

Anti-FAM3C antibody produced in rabbit is suitable for western blotting at a concentration of 1.0μg/ml.

Biochem/physiol Actions

FAM3C belongs to the family with sequence similarity 3 (FAM3) family and is a secreted cytokine that promotes epithelial to mesenchymal transition, metastasis and tumor formation in the epithelial cells. The activity of FAM3C is essential for formation of retinal lamina and the development of retina.

Sequence

Synthetic peptide located within the following region: DNWVFCGGKGIKTKSPFEQHIKNNKDTNKYEGWPEVVEMEGCIPQKQD

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

William S Streitfeld et al.
Cancer biology & therapy, 24(1), 2271638-2271638 (2023-11-06)
The poly(rC) binding protein 1 gene (PCBP1) encodes the heterogeneous nuclear ribonucleoprotein E1 (hnRNPE1), a nucleic acid-binding protein that plays a tumor-suppressive role in the mammary epithelium by regulating phenotypic plasticity and cell fate. Following the loss of PCBP1 function
Tatsuya Katahira et al.
Biochemical and biophysical research communications, 392(3), 301-306 (2010-01-12)
FAM3C is a secreted factor, which is involved in the epithelial to mesenchymal transition. In transcriptome profiling of the mouse retina using microarray, we found that FAM3C is highly expressed in the retina. FAM3C is expressed in the ganglion cell
Thomas Waerner et al.
Cancer cell, 10(3), 227-239 (2006-09-09)
Erk/MAPK and TGFbeta signaling cause epithelial to mesenchymal transition (EMT) and metastasis in mouse mammary epithelial cells (EpH4) transformed with oncogenic Ras (EpRas). In trials to unravel underlying mechanisms, expression profiling for EMT-specific genes identified a secreted interleukin-related protein (ILEI)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service