Skip to Content
Merck
All Photos(2)

Key Documents

AV44297

Sigma-Aldrich

Anti-C1QB antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Complement component 1, q subcomponent, B chain

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$3,345.00

R$3,345.00


Check Cart for Availability


Select a Size

Change View
100 μL
R$3,345.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$3,345.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

27 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... C1QB(713)

Immunogen

Synthetic peptide directed towards the C terminal region of human C1QB

Application

Anti-C1QB antibody produced in rabbit is suitable for western blotting at a concentration of 0.5μg/ml and for immunohistochemistry of paraffin-embedded tissue sections at a concentration of 4-8μg/ml.

Biochem/physiol Actions

C1QB is the B-chain polypeptide of the C1Q component of the human serum complement system. C1Q initiates complement activation and additionally modulates cytokine production, dendritic cell maturation and T-cell and B-cell responses.[1] Mutation in gene encoding for C1QB is associated with systemic lupus erythematosus[2] while polymorphisms in the gene have been observed in schizophrenia.[3]

Sequence

Synthetic peptide located within the following region: AYNTFQVTTGGMVLKLEQGENVFLQATDKNSLLGMEGANSIFSGFLLFPD

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yousuke Higuchi et al.
Pediatric rheumatology online journal, 11(1), 41-41 (2013-10-29)
C1q deficiency is a rare disease that is associated with a high probability of developing systemic lupus erythematosus. We report a 4-year-old Japanese girl who presented with fever, facial erythema, joint pain, and oral ulceration. Complement deficiencies were suspected because
Yi Cai et al.
PloS one, 9(3), e92340-e92340 (2014-03-22)
Complement functions as an important host defense system and complement C5 and C7 have been implicated in immunopathology of tuberculosis. However, little is known about the role of other complement components in tuberculosis. Complement gene expression in peripheral blood mononuclear
Roksana Zakharyan et al.
BMC medical genetics, 12, 126-126 (2011-09-29)
Schizophrenia is a complex, multifactorial psychiatric disorder. Our previous findings indicated that altered functional activity of the complement system, a major mediator of the immune response, is implicated in the pathogenesis of schizophrenia. In order to explore whether these alterations

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service