Skip to Content
Merck
All Photos(3)

Key Documents

AV41446

Sigma-Aldrich

Anti-MUC1 (AB2) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-CD227, Anti-EMA, Anti-Mucin, Anti-Mucin 1, cell surface associated, Anti-PEM, Anti-PUM

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$2,017.00

R$2,017.00


Check Cart for Availability


Select a Size

Change View
100 μL
R$2,017.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$2,017.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

22 kDa

species reactivity

dog, human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MUC1(4582)

General description

MUC1 (Mucin 1) is O-glycosylated, carbohydrate rich, transmembrane glycoprotein. It has approximately 13 members in the family. It is localized on the apical surface of mucosal epithelia in the lung, eye, breast and stomach. It is overexpressed in several epithelial cancers, including pancreatic ductal adenocarcinoma. It is a heterodimer consisting of N-terminal (MUC1-N) and C-terminal (MUC1-C) subunits. N-terminal (MUC1-N) end contains tandem repeats (VNTR) which forms the mucin component.

Immunogen

Synthetic peptide directed towards the C terminal region of human MUC1

Application

Anti-MUC1 (AB2) antibody produced in rabbit is suitable for immunohistochemistry and western blot.

Biochem/physiol Actions

MUC1 (Mucin 1) plays a major role in the formation of epithelium lining of the gastrointestinal tract. It lubricates the gastrointestinal tract to protect the epithelia from oesophagus to the colon. It also lubricates the epithelia of some organs such as pancreas and gall bladder. It is directly involved in several signaling pathways as s a modulator that affects oncogenesis, motility and metastasis.

Sequence

Synthetic peptide located within the following region: RRKNYGQLDIFPARDTYHPMSEYPTYHTHGRYVPPSSTDRSPYEKVSAGN

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Irina I Tyuryaeva et al.
Scientific reports, 7, 40217-40217 (2017-01-17)
Antitumor GO peptides have been designed as dimerization inhibitors of prominent oncoprotein mucin 1. In this study we demonstrate that activity of GO peptides is independent of the level of cellular expression of mucin 1. Furthermore, these peptides prove to
A P Corfield et al.
Frontiers in bioscience : a journal and virtual library, 6, D1321-D1357 (2001-10-02)
Mucins form part of the dynamic, interactive mucosal defensive system active at the mucosal surface of the gastrointestinal tract. They are carbohydrate rich glycoproteins with unique molecular structure and chemical properties. The family of mucin (MUC) genes has 13 members
C J Reid et al.
Gut, 42(2), 220-226 (1998-04-16)
Mucin glycoproteins play a key role in the normal function of the epithelium lining the gastrointestinal tract. The expression of mucin genes, MUC 3, 4, 5AC, 5B, 6, 7, and 8 in human fetal tissues was examined to establish the
M Sahraei et al.
Oncogene, 31(47), 4935-4945 (2012-01-24)
Pancreatic ductal adenocarcinoma (PDA) has one of the worst prognoses of all cancers. Mucin 1 (MUC1), a transmembrane mucin glycoprotein, is a key modulator of several signaling pathways that affect oncogenesis, motility and metastasis. Its expression is known to be
Lili Zhang et al.
The Journal of pathology, 234(1), 60-73 (2014-05-20)
Cigarette smoke increases the risk of lung cancer by 20-fold and accounts for 87% of lung cancer deaths. In the normal airway, heavily O-glycosylated mucin-1 (MUC1) and adherens junctions (AJs) establish a structural barrier that protects the airway from infectious

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service