Skip to Content
Merck
All Photos(1)

Key Documents

AV40961

Sigma-Aldrich

Anti-SF4 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-DKFZp434E2216, Anti-F23858, Anti-RBP, Anti-Splicing factor 4

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$3,345.00

R$3,345.00


Estimated to ship onMay 21, 2025



Select a Size

Change View
100 μL
R$3,345.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$3,345.00


Estimated to ship onMay 21, 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

72 kDa

species reactivity

bovine, mouse, horse, rat, dog, guinea pig, human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SF4(57794)

Related Categories

General description

Splicing factor 4/SURP and G patch domain containing 1 (SF4, RBP, SUGP1) is closely related to SFRS14 (arginine/serine-rich splicing factor 4) which is a potential normalization protein for gene expression studies in HCV-induced hepatocellular carcinoma (HCC).

Specificity

Anti-SF4 polyclonal antibody reacts with canine, bovine, zebrafish, human, mouse, and rat splicing factor 4/SURP and G patch domain containing 1 proteins.

Immunogen

Synthetic peptide directed towards the N terminal region of human SF4

Application

Anti-SF4 polyclonal antibody is used to tag splicing factor 4/SURP and G patch domain containing 1 for detection and quantitation by Western blotting and in plasma by immunohistochemical (IHC) techniques. It is used as a probe to determine the roles of splicing factor 4/SURP and G patch domain containing 1 in spliceosome formation and function.

Biochem/physiol Actions

SF4 is a member of the SURP family of splicing factors.SF4 is a member of the SURP family of splicing factors.[supplied by OMIM].

Sequence

Synthetic peptide located within the following region: MSLKMDNRDVAGKANRWFGVAPPKSGKMNMNILHQEELIAQKKREIEAKM

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kazuhiro Fukumura et al.
Nature communications, 12(1), 4910-4910 (2021-08-15)
Human pre-mRNA introns vary in size from under fifty to over a million nucleotides. We searched for essential factors involved in the splicing of human short introns by screening siRNAs against 154 human nuclear proteins. The splicing activity was assayed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service