Skip to Content
Merck
All Photos(3)

Key Documents

AV40390

Sigma-Aldrich

Anti-ACO1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Aconitase 1, soluble

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$3,345.00

R$3,345.00


Estimated to ship onApril 14, 2025



Select a Size

Change View
100 μL
R$3,345.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$3,345.00


Estimated to ship onApril 14, 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

98 kDa

species reactivity

pig, rat, human, bovine

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ACO1(48)

General description

Aconitase-1 is a soluble/cytoplasmin, non-mitochondrial enzyme that catalyzes the stereo-specific isomerization of citrate to isocitrate.

Specificity

Anti-Aconitase-1 antibody recognizes human aconitase-1.

Immunogen

Synthetic peptide directed towards the N terminal region of human ACO1

Application

Anti-Aconitase-1 antibody is a rabbit IgG polyclonal antibody used to tag Aconitase-1 protein(s) for detection and quantitation by Western blotting and in cells and tissues by immunohistochemical (IHC) techniques.

Biochem/physiol Actions

ACO1, also known as iron regulatory element binding protein 1 (IREB1), is a cytosolic protein which binds to iron-responsive elements (IREs). It plays a central role in cellular iron homeostasis. It was also shown to have aconitase activity, and hence grouped with the aconitase family of enzymes.Aconitase 1, also known as iron regulatory element binding protein 1 (IREB1), is a cytosolic protein which binds to iron-responsive elements (IREs). IREs are stem-loop structures found in the 5′ UTR of ferritin mRNA, and in the 3′ UTR of transferrin receptor mRNA. The iron-induced binding to the IRE results in repression of translation of ferritin mRNA, and inhibition of degradation of the otherwise rapidly degrading transferrin receptor mRNA. Thus, IREB1 plays a central role in cellular iron homeostasis. It was also shown to have aconitase activity, and hence grouped with the aconitase family of enzymes.

Sequence

Synthetic peptide located within the following region: MSNPFAHLAEPLDPVQPGKKFFNLNKLEDSRYGRLPFSIRVLLEAAIRNC

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service