Skip to Content
Merck
All Photos(2)

Documents

AV39623

Sigma-Aldrich

Anti-CBLL1 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Cas-Br-M (murine) ecotropic retroviral transforming sequence-like 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

55 kDa

species reactivity

bovine, rat, human, mouse, horse, dog

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CBLL1(79872)

General description

CBLL1 (Cbl proto-oncogene-like 1, E3 ubiquitin protein ligase) is a cbl-like ubiquitin ligase of E-cadherin complex. It is expressed in the neurons of central nervous system.
Rabbit Anti-CBLL1 antibody recognizes human, mouse, rat, zebrafish, canine, and chicken CBLL1.

Immunogen

Synthetic peptide directed towards the C terminal region of human CBLL1

Application

Rabbit Anti-CBLL1 antibody is suitable for western blot applications at a concentration of 1.25μg/ml.

Biochem/physiol Actions

CBLL1 (Cbl proto-oncogene-like 1, E3 ubiquitin protein ligase) is an E-cadherin binding protein involved in the ubiquitination of E-cadherin and regulation of E-cadherin complex endocytosis. During ubiquitination, it directly binds to the cytoplasmic domain of E-cadherin. It has been reported in a study that CBLL1 may play a role in neuronal apoptosis and in LPS (Lipopolysaccharide) administration. CBLL1 also have been predicted as a regulator of cell proliferation.

Sequence

Synthetic peptide located within the following region: PFTQPGGMSPGIWPAPRGPPPPPRLQGPPSQTPLPGPHHPDQTRYRPYYQ

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maohong Cao et al.
Journal of molecular histology, 44(2), 135-145 (2012-11-20)
CBLL1 (Casitas B-lineage lymphoma-transforming sequence-like protein 1) also known as Hakai, was originally identified as an E3 ubiquitin-ligase for the E-cadherin complex. Recent data have provided evidences for novel biological functional role of CBLL1 during tumor progression and other diseases.
Maria-Dolores Fernandez-Garcia et al.
Journal of virology, 85(6), 2980-2989 (2010-12-31)
The ubiquitin ligase CBLL1 (also known as HAKAI) has been proposed to be a critical cellular factor exploited by West Nile virus (WNV) for productive infection. CBLL1 has emerged as a major hit in a recent RNA interference screen designed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service