Skip to Content
Merck
All Photos(3)

Key Documents

AV39172

Sigma-Aldrich

Anti-POGZ antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Pogo transposable element with ZNF domain

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$2,017.00

R$2,017.00


Check Cart for Availability


Select a Size

Change View
100 μL
R$2,017.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$2,017.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

155 kDa

species reactivity

rat, mouse, dog, bovine, rabbit, guinea pig, horse, human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... POGZ(23126)

General description

POGZ is a zinc-finger protein known to interact with HP1α. Studies have reported that POGZ regulates the dissociation of Aurora B kinase from chromosomal arms during the M phase of the cell cycle.
Rabbit Anti-POGZ antibody recognizes human, mouse, rat, canine, rabbit, and bovine POGZ.

Immunogen

Synthetic peptide directed towards the N terminal region of human POGZ

Application

Rabbit Anti-POGZ antibody is suitable for western blot (5.0 μg/ml) and IHC (4-8 μg/ml) applications.

Biochem/physiol Actions

POGZ appears to be a zinc finger protein containing a transposase domain at the C-terminus. This protein was found to interact with the transcription factor SP1 in a yeast two-hybrid systemThe protein encoded by this gene appears to be a zinc finger protein containing a transposase domain at the C-terminus. This protein was found to interact with the transcription factor SP1 in a yeast two-hybrid system. At least three alternatively spliced transcript variants encoding distinct isoforms have been observed.

Sequence

Synthetic peptide located within the following region: EELEPWQKISDVIEDSVVEDYNSVDKTTTVSVSQQPVSAPVPIAAHASVA

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ryu-Suke Nozawa et al.
Nature cell biology, 12(7), 719-727 (2010-06-22)
Heterochromatin protein 1 (HP1) has an essential role in heterochromatin formation and mitotic progression through its interaction with various proteins. We have identified a unique HP1alpha-binding protein, POGZ (pogo transposable element-derived protein with zinc finger domain), using an advanced proteomics
Kensuke Matsumura et al.
Nature communications, 11(1), 859-859 (2020-02-28)
Pogo transposable element derived with ZNF domain (POGZ) has been identified as one of the most recurrently de novo mutated genes in patients with neurodevelopmental disorders (NDDs), including autism spectrum disorder (ASD), intellectual disability and White-Sutton syndrome; however, the neurobiological

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service