Skip to Content
Merck
All Photos(2)

Key Documents

AV38287

Sigma-Aldrich

Anti-NR2C1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Nuclear receptor subfamily 2, group C, member 1, Anti-TR2, Anti-TR2-11

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$3,345.00

R$3,345.00


Check Cart for Availability


Select a Size

Change View
100 μL
R$3,345.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$3,345.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

67 kDa

species reactivity

guinea pig, rat, horse, bovine, human, mouse, dog

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NR2C1(7181)

General description

Testicular receptor 2 /nuclear receptor subfamily 2, group C, member 1 (NR2C1, TR2) is a nuclear receptor transcription factor. TR2 is a testicular orphan nuclear receptors that acts to regulate other nuclear receptors. TR2 is believed to regulate early embryonic development by regulating key genes involved in stem cell self-renewal, commitment and differentiation. TR2/TR4 directly represses Gata1/GATA1 transcription in murine and human erythroid progenitor cells.

The previously assigned protein identifier Q15625 has been merged into P13056. Full details can be found on the UniProt database.

Specificity

Anti-NR2C1 polyclonal antibody reacts with zebrafish, canine, bovine, human, mouse, and rat testicular receptor 2 proteins.

Immunogen

Synthetic peptide directed towards the C terminal region of human NR2C1

Application

Anti-NR2C1 polyclonal antibody is used to tag testicular receptor 2 /Nuclear receptor subfamily 2, group C, member 1 protein for detection and quantitation by Western blotting and in plasma by immunohistochemical (IHC) techniques. It is used as a probe to determine the roles of testicular receptor 2 in early embryonic development and stem cell self-renewal, commitment and differentiation.

Biochem/physiol Actions

The nuclear orphan receptors NR2C1 represses transcription and binds DNA as a homodimer. NR2C1 binds the IR7 element in the promoter of its own gene in an autoregulatory negative feedback mechanism. NR2C1 may function as a negative modulator to suppress androgen receptor function in prostate cancer. NR2C1 may exert an important repressor in regulating ER activity in mammary glands. The nuclear orphan receptors TR2 (NR2C1) and TR4 form a heterodimer that binds to the epsilon and gamma globin promoter DR1 sites

Sequence

Synthetic peptide located within the following region: LPALRLMNATITEELFFKGLIGNIRIDSVIPHILKMEPADYNSQIIGHSI

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service