Skip to Content
Merck
All Photos(3)

Key Documents

AV38232

Sigma-Aldrich

Anti-SOX2 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-ANOP3, Anti-MGC2413, Anti-SRY (sex determining region Y)-box 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$2,017.00

R$2,017.00


Estimated to ship onMay 29, 2025



Select a Size

Change View
100 μL
R$2,017.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$2,017.00


Estimated to ship onMay 29, 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

34 kDa

species reactivity

human, rat, bovine, goat, pig, mouse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... SOX2(6657)

Immunogen

Synthetic peptide directed towards the N terminal region of human SOX2

Biochem/physiol Actions

SOX2 is a member of the SRY-related HMG-box (SOX) family of transcription factors involved in the regulation of embryonic development and in the determination of cell fate. The protein may act as a transcriptional activator after forming a protein complex with other proteins. Mutations in this gene have been associated with bilateral anophthalmia, a severe form of structural eye malformation.This intronless gene encodes a member of the SRY-related HMG-box (SOX) family of transcription factors involved in the regulation of embryonic development and in the determination of cell fate. The encoded protein may act as a transcriptional activator after forming a protein complex with other proteins. Mutations in this gene have been associated with bilateral anophthalmia, a severe form of structural eye malformation. This gene lies within an intron of another gene called SOX2 overlapping transcript (SOX2OT).

Sequence

Synthetic peptide located within the following region: AAGGNQKNSPDRVKRPMNAFMVWSRGQRRKMAQENPKMHNSEISKRLGAE

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anran Fan et al.
Reproduction (Cambridge, England), 146(6), 569-579 (2013-09-21)
TET1 is implicated in maintaining the pluripotency of embryonic stem cells. However, its precise effects on induced pluripotent stem cells (iPSCs), and particularly on porcine iPSCs (piPSCs), are not well defined. To investigate the role of TET1 in the pluripotency
Lina Tang et al.
Cellular reprogramming, 14(4), 342-352 (2012-07-11)
Direct reprogramming of somatic cells to induced pluripotent stem cells (iPSCs) provides an invaluable resource for regenerative medicine. Because of some ethical and logistical barriers, human iPSCs cannot be used to generate a chimera, which is one of markers representing
Irem Dogan et al.
Lung cancer (Amsterdam, Netherlands), 85(1), 1-6 (2014-04-22)
Primary and acquired resistance to EGFR TKIs in EGFR mutant lung cancer occurs primarily through secondary mutations in EGFR or Met amplification. Drug resistance can also be mediated by expression of pluripotency transcription factors, such as OCT4, SOX2 and NANOG
Xiangxin Lou et al.
Neuroscience letters, 579, 1-6 (2014-07-13)
The presence of stem cells in the organ of Corti raises the hope of regeneration of mammalian inner ear cells. However, little is known about the distribution of endogenous stem cells in the inner ear as well as their sphere-forming
Ana Benito-Gonzalez et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(38), 12865-12876 (2014-09-19)
Mechano-sensory hair cells (HCs), housed in the inner ear cochlea, are critical for the perception of sound. In the mammalian cochlea, differentiation of HCs occurs in a striking basal-to-apical and medial-to-lateral gradient, which is thought to ensure correct patterning and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service