Skip to Content
Merck
All Photos(3)

Key Documents

AV35307

Sigma-Aldrich

Anti-MCOLN1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Mucolipin 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$3,345.00

R$3,345.00


Check Cart for Availability


Select a Size

Change View
100 μL
R$3,345.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$3,345.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

64 kDa

species reactivity

pig, rabbit, dog, human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MCOLN1(57192)

General description

MCOLN1 codes for a transmembrane protein that is found in vesicles. It is involved in endocytosis and lysosomal exocytosis. MCOLN1 mutations have been associated with the neurodegenerative lysosomal storage disorder, mucolipidosis type IV (MLIV).
Rabbit Anti-MCOLN1 antibody recognizes bovine, canine, human, rat, and mouse MCOLN1.

Immunogen

Synthetic peptide directed towards the N terminal region of human MCOLN1

Application

Rabbit Anti-MCOLN1 antibody is suitable for western blot applications at a concentration of 0.5 μg/ml.

Biochem/physiol Actions

MCOLN1 encodes a protein that may be involved in calcium signaling and membrane trafficking in mucolipidosis IV.

Sequence

Synthetic peptide located within the following region: FRHLFLLGYSDGADDTFAAYTREQLYQAIFHAVDQYLALPDVSLGRYAYV

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gideon Bach et al.
Human mutation, 26(6), 591-591 (2005-11-16)
Mucolipidosis type IV (MLIV) is a neurodegenerative lysosomal storage disorder that occurs in an increased frequency in the Ashkenazi Jewish (AJ) population. The frequency of the disease in this population has been established by the testing of 66,749 AJ subjects
Yin Xu et al.
Molecular psychiatry, 26(10), 5925-5939 (2020-05-06)
Neurofibrillary tangles (NFTs) composed of hyperphosphorylated and misfolded tau protein are a pathological hallmark of Alzheimer's disease and other tauopathy conditions. Tau is predominantly an intraneuronal protein but is also secreted in physiological and pathological conditions. The extracellular tau has

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service