Skip to Content
Merck
All Photos(1)

Key Documents

AV34760

Sigma-Aldrich

Anti-PHF17 (AB2) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-FLJ22479, Anti-JADE1, Anti-KIAA1807, Anti-PHD finger protein 17

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$3,345.00

R$3,345.00


Check Cart for Availability


Select a Size

Change View
100 μL
R$3,345.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$3,345.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

58 kDa

species reactivity

bovine, rat, guinea pig, horse, human, mouse, rabbit, dog

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PHF17(79960)

General description

PHF17 (PHD finger protein 17) is a short-lived, kidney-enriched Jade protein consisting of canonical plant homeodomain (PHD) finger protein and a non-canonical extended PHD with zinc-binding ability. It is localized in the nucleus and is highly expressed in kidney and renal proximal tubule cells. Rabbit Anti-PHF17 antibody recognizes human, mouse, rat, bovine, and chicken PHF17.

Immunogen

Synthetic peptide directed towards the C terminal region of human PHF17

Application

Rabbit Anti-PHF17 antibody is suitable for western blot applications at a concentration of 1μg/ml.
Rabbit Anti-PHF17 antibody is suitable for western blot applications.

Biochem/physiol Actions

PHF17 (PHD finger protein 17) is associated with several cellular activities such as chromatin remodeling, renal tubular epithelial cell differentiation, growth suppression, apoptosis and protein-protein interactions. It possesses transcriptional and endogenous histone acetyltransferase (HAT) activity. It acts as a transcriptional co-activator in the TIP60 mediated histone H4/H2A specific HAT activity. It has been reported that PHF17 may play a role in the renal cancer and von Hippel-Lindau disease. PHF17 also possesses tumour suppressor property. In the renal tumorigenesis, it controls canonical Wnt signaling pathway by ubiquitylating oncoprotein β-catenin in Wnt-responsive manner.

Sequence

Synthetic peptide located within the following region: EPFASLEQNREEAHRVSVRKQKLQQLEDEFYTFVNLLDVARALRLPEEVV

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria V Panchenko et al.
The Journal of biological chemistry, 279(53), 56032-56041 (2004-10-27)
Jade-1 was identified as a protein partner of the von Hippel-Lindau tumor suppressor pVHL. The interaction of Jade-1 and pVHL correlates with renal cancer risk. We have investigated the molecular function of Jade-1. Jade-1 has two zinc finger motifs called
Vipul C Chitalia et al.
Nature cell biology, 10(10), 1208-1216 (2008-09-23)
The von Hippel-Lindau protein pVHL suppresses renal tumorigenesis in part by promoting the degradation of hypoxia-inducible HIF-alpha transcription factors; additional mechanisms have been proposed. pVHL also stabilizes the plant homeodomain protein Jade-1, which is a candidate renal tumour suppressor that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service