Skip to Content
Merck
All Photos(1)

Key Documents

AV34680

Sigma-Aldrich

Anti-MID1 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Midline 1 (Opitz/BBB syndrome)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$2,577.00

R$2,577.00


Check Cart for Availability


Select a Size

Change View
100 μL
R$2,577.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$2,577.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

75 kDa

species reactivity

human, bovine, guinea pig, dog, rat, mouse, horse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MID1(4281)

Immunogen

Synthetic peptide directed towards the C terminal region of human MID1

Biochem/physiol Actions

MID1 is a member of the tripartite motif (TRIM) family, also known as the ′RING-B box-coiled coil′ (RBCC) subgroup of RING finger proteins. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. This protein forms homodimers which associate with microtubules in the cytoplasm. The protein is likely involved in the formation of multiprotein structures acting as anchor points to microtubules. Mutations in this gene have been associated with the X-linked form of Opitz syndrome, which is characterized by midline abnormalities such as cleft lip, laryngeal cleft, heart defects, hypospadias, and agenesis of the corpus callosum. This gene was also the first example of a gene subject to X inactivation in human while escaping it in mouse. Several different transcript variants are generated by alternate splicing; however, the full length nature of two variants has not been determined.

Sequence

Synthetic peptide located within the following region: AINQAGSRSSEPGKLKTNSQPFKLDPKSAHRKLKVSHDNLTVERDESSSK

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service