Skip to Content
Merck
All Photos(3)

Key Documents

AV34382

Sigma-Aldrich

Anti-NFKBIB antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, β

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$3,042.00

R$3,042.00


Check Cart for Availability


Select a Size

Change View
100 μL
R$3,042.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$3,042.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

38 kDa

species reactivity

rabbit, mouse, human, rat, guinea pig

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... NFKBIB(4793)

General description

NFKBIB is a NF-κB inhibitor that forms a complex with NF-κB and traps it in the cytoplasm. Serine phosphorylation in NFKBIB proteins activates its ubiquitin-mediated degradation. This subsequently facilitates the nuclear transport and functions of NF-κB proteins.
Rabbit Anti-NFKBIB antibody recognizes human, mouse, rat, canine, and bovine NFKBIB.

Immunogen

Synthetic peptide directed towards the N terminal region of human NFKBIB

Application

Rabbit Anti-NFKBIB antibody can be used for western blot applications at a concentration of 0.5 μg/ml.

Biochem/physiol Actions

NFKB1 (MIM 164011) or NFKB2 (MIM 164012) is bound to REL (MIM 164910), RELA (MIM 164014), or RELB (MIM 604758) to form the NFKB complex. The NFKB complex is inhibited by I-kappa-B proteins (NFKBIA, MIM 164008, or NFKBIB), which inactivate NF-kappa-B by trapping it in the cytoplasm. Phosphorylation of serine residues on the I-kappa-B proteins by kinases (IKBKA, MIM 600664 or IKBKB, MIM 603258) marks them for destruction via the ubiquitination pathway, thereby allowing activation of the NF-kappa-B complex. Activated NFKB complex translocates into the nucleus and binds DNA at kappa-B-binding motifs such as 5-prime GGGRNNYYCC 3-prime or 5-prime HGGARNYYCC 3-prime (where H is A, C, or T; R is an A or G purine; and Y is a C or T pyrimidine).[supplied by OMIM].

Sequence

Synthetic peptide located within the following region: LVFGYVTEDGDTALHLAVIHQHEPFLDFLLGFSAGTEYMDLQNDLGQTAL

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shanyu Zhao et al.
PloS one, 9(7), e102273-e102273 (2014-07-16)
Prenatal exposure to Lipopolysaccharide (LPS) produces hypertension in adult offspring rats. The present study was to explore the effects of prenatal inflammation on morphological and functional changes in the aorta from offspring rats and to further assess its susceptibility to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service