Skip to Content
Merck
All Photos(1)

Key Documents

AV34285

Sigma-Aldrich

Anti-CORO1A (AB1) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Coronin, actin binding protein, 1A

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$2,017.00

R$2,017.00


Check Cart for Availability


Select a Size

Change View
100 μL
R$2,017.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$2,017.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

51 kDa

species reactivity

guinea pig, human, dog, bovine, rat, horse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CORO1A(11151)

General description

CORO1A is a WD-repeat protein that has been implicated in Mycobacterium leprae infection. Studies have reported that CORO1A localizes on the membrane of phagosomes that contain M. leprae, where Toll-like receptor (TLR) 2 is also present. CORO1A suppresses TLR-mediated signaling in human macrophages.
Rabbit Anti-CORO1A recognizes bovine, rat, rabbit, mouse, human, and canine CORO1A.

Immunogen

Synthetic peptide directed towards the C terminal region of human CORO1A

Application

Rabbit Anti-CORO1A can be used for western blot applications at a concentration of 1.25 μg/ml.

Biochem/physiol Actions

CORO1A forms homodimers. It plays a role in the cross-linking of F-actin in the cell.

Sequence

Synthetic peptide located within the following region: TGRRRAAPEASGTPSSDAVSRLEEEMRKLQATVQELQKRLDRLEETVQAK

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Koichi Suzuki et al.
Acta histochemica et cytochemica, 39(4), 107-112 (2007-03-01)
Mycobacteria have acquired an intracellular lifestyle within the macrophage, which is best exemplified by the enlarged infected histiocytes seen in lepromatous leprosy. To survive within the cell, mycobacteria must escape intracellular bactericidal mechanisms. In a study of Mycobacterium bovis Bacille
K Tanigawa et al.
Clinical and experimental immunology, 156(3), 495-501 (2009-05-15)
Mycobacterium leprae is an intracellular pathogen that survives within the phagosome of host macrophages. Several host factors are involved in producing tolerance, while others are responsible for killing the mycobacterium. Tryptophan aspartate-containing coat protein (TACO; also known as CORO1A or

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service