Skip to Content
Merck
All Photos(1)

Documents

AV32810

Sigma-Aldrich

Anti-KLF1 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Kruppel-like factor 1 (erythroid)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

38 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KLF1(10661)

General description

Kruppel-like factor 1 (erythroid) is a transcription factor required for the proper differentiation of erythroid (red blood) cells from bipotent progenitor cells. Kruppel-like factor 1 regulates erythroid cell differenation and maturation via the processes of transcriptional activation, gamma to β globin switching and chromatin remodeling.
Rabbit polyclonal anti-KLF1 antibody reacts with pig, human, and bovine Kruppel-like factor 1 (erythroid) transcription factors.

Immunogen

Synthetic peptide directed towards the middle region of human KLF1

Application

Rabbit Anti-KLF1 antibody can be used for western blot applications at a concentration of 2.5μg/ml.
Rabbit polyclonal anti-KLF1 antibody is used to tag Kruppel-like factor 1 (erythroid) for detection and quantitation by immunocytochemical and immunohistochemical (IHC) techniques. It is used as a probe to determine the presence and roles of Kruppel-like factor 1 (erythroid) in erythroid cell differentiation and maturation.

Biochem/physiol Actions

KLF1 is a transcription factor, originally identified in this laboratory, which plays a crucial role as a transcriptional activator at the adult beta-globin locus.

Sequence

Synthetic peptide located within the following region: SVPAASGAPYGLLSGYPAMYPAPQYQGHFQLFRGLQGPAPGPATSPSFLS

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service