Skip to Content
Merck
All Photos(3)

Key Documents

AV32738

Sigma-Aldrich

Anti-MyEF2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Myelin expression factor 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$3,345.00

R$3,345.00


Estimated to ship onApril 21, 2025



Select a Size

Change View
100 μL
R$3,345.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$3,345.00


Estimated to ship onApril 21, 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

64 kDa

species reactivity

dog, rat, human, rabbit, mouse, bovine, horse, guinea pig

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MYEF2(50804)

General description

Myelin basic protein (MBP) is a major component of the myelin sheath whose production is developmentally controlled during myelinogenesis. Myelin expression factor 2 (MyEF2), a single-stranded DNA binding factor, is a repressor of myelin basic protein (MBP) expression. MyEF2 interacts with RUNX1 to represses hematopoietic genes in erythroid cells.
Rabbit polyclonal anti-MyEF2 antibody reacts with bovine, canine, human, mouse, rat, zebrafish, and chicken myelin expression factor 2 proteins.

Immunogen

Synthetic peptide directed towards the middle region of human MYEF2

Application

Rabbit Anti-MyEF2 can be used for western blot (1 μg/ml) and IHC (4-8 μg/ml) applications.
Rabbit polyclonal anti-MyEF2 antibody is used to tag myelin expression factor 2 for detection and quantitation by immunocytochemical and immunohistochemical (IHC) techniques. It is used as a probe to determine the presence and roles of myelin expression factor 2 in the repression of genes involved in myelinogenesis and hematopoiesis.

Biochem/physiol Actions

MEF-2 is expressed early in the differentiation program and is suppressed by specific polypeptide growth factors. The ability of MEF-2 to recognize conserved activating elements associated with multiple-specific genes suggests that this factor may participate in the coordinate regulation of genes during myogenesis.

Sequence

Synthetic peptide located within the following region: QAGRLGSTIFVANLDFKVGWKKLKEVFSIAGTVKRADIKEDKDGKSRGMG

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V Muralidharan et al.
Journal of cellular biochemistry, 66(4), 524-531 (1997-09-15)
Myelin basic protein (MBP) is a major component of the myelin sheath whose production is developmentally controlled during myelinogenesis. Earlier studies have indicated that programmed expression of the MBP gene is regulated at the level of transcription. Evidently, the MB1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service