Skip to Content
Merck
All Photos(1)

Key Documents

AV32623

Sigma-Aldrich

Anti-HIC1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Hypermethylated in cancer 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$3,345.00

R$3,345.00


Check Cart for Availability


Select a Size

Change View
100 μL
R$3,345.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

R$3,345.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

74 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... HIC1(3090)

General description

HIC1 is a tumor suppressor that interacts with SIRT1 to regulate p53-depenedent responses to DNA damage[1].
Rabbit Anti-HIC1 antibody recognizes human, mouse, and rat HIC1.

Immunogen

Synthetic peptide directed towards the N terminal region of human HIC1

Application

Rabbit Anti-HIC1 antibody can be used for western blot applications at a concentration of 0.25μg/ml.

Biochem/physiol Actions

HIon Channel1 is a transcriptional repressor. HIon Channel1 may act as a tumor suppressor. HIon Channel1 may be involved in development of head, face, limbs and ventral body wall. Defects in HIon Channel1 may be a cause of the facial dysmorphism and additional birth defects (except for lissencephaly) seen in the contiguous gene disorder Miller-Dieker syndrome (MDS), like defective development of the nose, jaws, extremities, gastrointestinal tract, and kidney.

Sequence

Synthetic peptide located within the following region: ALCASERRCSPLCGLDLSKKSPPGSAAPERPLAERELPPRPDSPPSAGPA

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wen Yong Chen et al.
Cell, 123(3), 437-448 (2005-11-05)
Hypermethylated in cancer 1 (HIC1) is an epigenetically regulated transcriptional repressor that functionally cooperates with p53 to suppress age-dependent development of cancer in mice. Here we show that the mechanism by which the loss of HIC1 function promotes tumorigenesis is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service