Skip to Content
Merck
All Photos(2)

Documents

AV32053

Sigma-Aldrich

Anti-NPAS1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Neuronal PAS domain protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

63 kDa

species reactivity

rabbit, human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NPAS1(4861)

Related Categories

General description

NPAS1 is a basic helix-loop-helix transcription factor that that regulates branching morphogenesis in mouse embryonic lung tissues. Studies in mice have reported that NPAS1 deficiency can result in behavioural and functional abnormalities.
Rabbit Anti-NPAS1 antibody recognizes human, mouse, and rat NPAS1.

Immunogen

Synthetic peptide directed towards the C terminal region of human NPAS1

Application

Rabbit Anti-NPAS1 antibody can be used for western blotting at 2μg/ml. It can also be used for IHC at 4-8μg/ml, using paraffin-embedded tissues.

Biochem/physiol Actions

NPAS1 is a member of the basic helix-loop-helix (bHLH)-PAS family of transcription factors. Studies of a related mouse gene suggest that it functions in neurons. The exact function of this gene is unclear, but it may play protective or modulatory roles during late embryogenesis and postnatal development.

Sequence

Synthetic peptide located within the following region: TIRYGPAELGLVYPHLQRLGPGPALPEAFYPPLGLPYPGPAGTRLPRKGD

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Claudia Erbel-Sieler et al.
Proceedings of the National Academy of Sciences of the United States of America, 101(37), 13648-13653 (2004-09-07)
Laboratory mice bearing inactivating mutations in the genes encoding the NPAS1 and NPAS3 transcription factors have been shown to exhibit a spectrum of behavioral and neurochemical abnormalities. Behavioral abnormalities included diminished startle response, as measured by prepulse inhibition, and impaired
Bernadette M Levesque et al.
American journal of respiratory cell and molecular biology, 36(4), 427-434 (2006-11-18)
Drosophila trachealess (Trl), master regulator of tracheogenesis, has no known functional mammalian homolog. We hypothesized that genes similar to trachealess regulate lung development. Quantitative (Q)RT-PCR and immunostaining were used to determine spatial and temporal patterns of npas1 gene expression in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service