Skip to Content
Merck
All Photos(2)

Key Documents

AV31694

Sigma-Aldrich

Anti-FOXF2 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Forkhead box F2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$2,017.00

R$2,017.00


Estimated to ship onMay 28, 2025



Select a Size

Change View
100 μL
R$2,017.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$2,017.00


Estimated to ship onMay 28, 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

46 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FOXF2(2295)

General description

FoxF2 is a forkhead transcription factor that is expressed in mesodermal tissues. This transcription factor is involved in epithelio-mesenchymal crosstalk. It may also function as an independent regulator of development and palatogenesis.
Rabbit Anti-FOXF2 recognizes human FOXF2.

Immunogen

Synthetic peptide directed towards the N terminal region of human FOXF2

Application

Rabbit Anti-FOXF2 antibody can be used for western blot assays at a concentration of 1μg/ml.

Biochem/physiol Actions

FOXF2 encodes forkhead box F2, one of many human homologues of the Drosophila melanogaster transcription factor forkhead. FOXF2 is expressed in lung and placenta, and has been shown to transcriptionally activate several lung-specific genes.

Sequence

Synthetic peptide located within the following region: SSASCASSSSSSNSASAPSAACKSAGGGGAGAGSGGAKKASSGLRRPEKP

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Aitola et al.
Developmental dynamics : an official publication of the American Association of Anatomists, 218(1), 136-149 (2000-05-24)
The growing family of forkhead transcription factors plays many important roles during embryonic development. In this study we have used in situ hybridization to explore the expression pattern of the forkhead transcription factor gene FoxF2 (FREAC-2, LUN) during mouse and
Tao Wang et al.
Developmental biology, 259(1), 83-94 (2003-06-19)
The forkhead genes encode a transcription factor involved in embryogenesis and pattern formation in multicellular organisms. They are mammalian transcriptional regulators that bind DNA as a monomer through their forkhead domain. The Foxf2 (LUN) mRNA is expressed in the mesenchyme

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service