Skip to Content
Merck
All Photos(1)

Key Documents

AV31526

Sigma-Aldrich

Anti-SUPT16H antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-CDC68, Anti-FACT, Anti-FACTP140, Anti-FLJ10857, Anti-FLJ14010, Anti-FLJ34357, Anti-SPT16/CDC68, Anti-Suppressor of Ty 16 homolog (S. cerevisiae)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$2,017.00

R$2,017.00


Check Cart for Availability


Select a Size

Change View
100 μL
R$2,017.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$2,017.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

66 kDa

species reactivity

rabbit, rat, human, pig, horse, dog

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SUPT16H(11198)

General description

SUPT16H is a transcription factor that interacts with RNF40. This transcription factor mediates structural changes in chromatin during DNA double strand repair.
Rabbit Anti-SUPT16H antibody recognizes bovine, rat, human, zebrafish, mouse, and canine SUPT16H.

Immunogen

Synthetic peptide directed towards the N terminal region of human SUPT16H

Application

Rabbit Anti-SUPT16H antibody can be used for western blot assays at a concentration of 1.25μg/ml.

Biochem/physiol Actions

Transcription of protein-coding genes can be reconstituted on naked DNA with only the general transcription factors and RNA polymerase II. However, this minimal system cannot transcribe DNA packaged into chromatin, indicating that accessory factors may facilitate access to DNA. One such factor, FACT (facilitates chromatin transcription), interacts specifically with histones H2A/H2B to effect nucleosome disassembly and transcription elongation. FACT is composed of an 80 kDa subunit and a 140 kDa subunit, the latter of which is SUPT16H.

Sequence

Synthetic peptide located within the following region: ASKKKVEFLKQIANTKGNENANGAPAITLLIREKNESNKSSFDKMIEAIK

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vijayalakshmi Kari et al.
Cell cycle (Georgetown, Tex.), 10(20), 3495-3504 (2011-10-28)
Many anticancer therapies function largely by inducing DNA double-strand breaks (DSBs) or altering the ability of cancer cells to repair them. Proper and timely DNA repair requires dynamic changes in chromatin assembly and disassembly characterized by histone H3 lysine 56

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service