Skip to Content
Merck
All Photos(4)

Key Documents

AV31443

Sigma-Aldrich

Anti-HHEX antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Hematopoietically expressed homeobox

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$3,345.00

R$3,345.00


Estimated to ship onMay 30, 2025



Select a Size

Change View
100 μL
R$3,345.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$3,345.00


Estimated to ship onMay 30, 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

30 kDa

species reactivity

human, horse, rat, dog

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HHEX(3087)

General description

HHEX is a homeobox protein that is involved in developmental functions. HHEX variations have been identified as risk factors for diabetes type 2 in Japanese population groups.
Rabbit Anti-HHEX antibody recognizes pig, human, canine, bovine, mouse, and rat HHEX.

Immunogen

Synthetic peptide directed towards the middle region of human HHEX

Application

Rabbit Anti-HHEX antibody is suitable for use in western blot (0.5μg/ml) and immunohistochemistry (4-8μg/ml) applications.

Biochem/physiol Actions

HHEX encodes a member of the homeobox family of transcription factors, many of which are involved in developmental processes. Expression in specific hematopoietic lineages suggests that this protein may play a role in hematopoietic differentiation.

Sequence

Synthetic peptide located within the following region: KQENPQSNKKEELESLDSSCDQRQDLPSEQNKGASLDSSQCSPSPASQED

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Horikoshi et al.
Diabetologia, 50(12), 2461-2466 (2007-10-12)
Recently, several groups have carried out whole-genome association studies in European and European-origin populations and found novel type 2 diabetes-susceptibility genes, fat mass and obesity associated (FTO), solute carrier family 30 (zinc transporter), member 8 (SLC30A8), haematopoietically expressed homeobox (HHEX)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service