Skip to Content
Merck
All Photos(2)

Key Documents

AV31422

Sigma-Aldrich

Anti-SPIB antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Spi-B transcription factor (Spi-1/PU.1 related)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$3,345.00

R$3,345.00


Check Cart for Availability


Select a Size

Change View
100 μL
R$3,345.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$3,345.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

19 kDa

species reactivity

bovine, dog, rabbit, guinea pig, human, rat, horse, mouse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SPIB(6689)

General description

SPIB is a transcriptional stimulator that associates with PU-box and functions as a lymphoid enhancer. SPIB variants have been linked to primary biliary cirrhosis.
Rabbit Anti-SPIB antibody recognizes zebrafish, canine, pig, bovine, chicken, human, mouse, and rat SPIB.

Immunogen

Synthetic peptide directed towards the C terminal region of human SPIB

Application

Rabbit Anti-SPIB antibody can be used for western blot applications at 0.25μg/ml.

Biochem/physiol Actions

SPI1 (MIM 165170) and SPIB are members of a subfamily of ETS (see ETS1; MIM 164720) transcription factors. ETS proteins share a conserved ETS domain that mediates specific DNA binding. SPIB and SPI1 bind to a purine-rich sequence, the PU box (5-prime-GAGGAA-3-prime).

Sequence

Synthetic peptide located within the following region: GQQKGNRKRMTYQKLARALRNYAKTGEIRKVKRKLTYQFDSALLPAVRRA

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G M Hirschfield et al.
Genes and immunity, 13(4), 328-335 (2012-01-20)
We fine mapped two primary biliary cirrhosis (PBC) risk loci, CLEC16A (C-type lectin domain family 16 member A)-suppressor of cytokine signaling 1 (SOCS1) and Spi-B protein (SPIB) and sequenced a locus, sialic acid acetylesterase (SIAE), proposed to harbor autoimmunity-associated mutations.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service