Skip to Content
Merck
All Photos(4)

Key Documents

AV31375

Sigma-Aldrich

Anti-EN2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-AUTS1, Anti-AUTS10, Anti-Engrailed homeobox 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$3,345.00

R$3,345.00


Check Cart for Availability


Select a Size

Change View
100 μL
R$3,345.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

R$3,345.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

34 kDa

species reactivity

dog, mouse, guinea pig, horse, human, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... EN2(2020)

General description

Engrailed homeodomain-containing transcription factors play crucial roles in brain development across many species, including the determination of the hindbrain/midbrain border, cerebellar patterning and aiding in neuronal axon guidance. Engrailed-2 (En-2) gene is expressed across the mesencephalon/metencephalon (mes/met) boundary in the cerebellar primordium. Engrailed-2 is involved in the determination of skeletal muscle physiologic properties. Urinary EN2 is a highly specific and sensitive candidate biomarker of prostate cancer.
Rabbit polyclonal anti-ENS antibody reacts with chicken, zebrafish, human, mouse, and rat engrailed homeobox 2 transcription factors.

Immunogen

Synthetic peptide directed towards the C-terminal region of Human EN2

Application

Rabbit Anti-EN2 antibody is suitable for use in western blot (0.5μg/ml) assays.
Rabbit polyclonal anti-ENS antibody is used to tag engrailed homeobox 2 transcription factor for detection and quantitation by immunocytochemical and immunohistochemical (IHC) techniques. It is used as a probe to determine the presence and roles of engrailed homeobox 2 transcription factor in brain development and skeletal muscle differentiation.

Biochem/physiol Actions

Homeobox-containing genes are thought to have a role in controlling development. The human engrailed homologs 1 and 2 encode homeodomain-containing proteins and have been implicated in the control of pattern formation during development of the central nervous system.

Sequence

Synthetic peptide located within the following region: NESQIKIWFQNKRAKIKKATGNKNTLAVHLMAQGLYNHSTTAKEGKSDSE

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service