Skip to Content
Merck
All Photos(2)

Key Documents

AV31200

Sigma-Aldrich

Anti-ISGF3G antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-IRF-9, Anti-ISGF3, Anti-ISGF3G, Anti-p48

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$3,345.00

R$3,345.00


Estimated to ship onMay 19, 2025



Select a Size

Change View
100 μL
R$3,345.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$3,345.00


Estimated to ship onMay 19, 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

44 kDa

species reactivity

rabbit, human, rat, bovine, mouse, guinea pig, dog, horse

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... ISGF3G(10379)

Related Categories

General description

Interferon regulatory factor 9 (ISGF3G, IRF-9, p48) is a DNA-binding component of the heterotrimeric transcription factor ISGF3 which is activated by interferon-α and β (type I IFNs). The other two components of ISGF3 are STAT1 and STAT2. ISGF3 gamma or complexes containing ISGF3 gamma are involved in IRF-1-independent pathways mediating IFN gene regulation. ISGF3 plays a primary role in the transmission of a signal from the cell surface to the nucleus via regulatory factors p84/91 and p113.
Rabbit polyclonal anti-ISGF3G antibody reacts with pig, bovine, human, mouse, rat, and zebrafish interferon regulatory factor 9/ISGF3 gamma proteins.

Immunogen

Synthetic peptide directed towards the N terminal region of human ISGF3G

Application

Rabbit Anti-ISGF3G antibody can be used for western blotting applications at 2.5μg/ml.
Rabbit polyclonal anti-ISGF3G antibody is used to tag interferon regulatory factor 9/ISGF3 gamma for detection and quantitation by immunocytochemical and immunohistochemical (IHC) techniques. It is used as a probe to determine the presence and roles of interferon regulatory factor 9/ISGF3 gamma in the mediation of interferon-α and β (type I IFNs) signaling.

Biochem/physiol Actions

ISGF3G functions to recruit RNA polymerase II to the promoter of interferon-stimulated genes and requires histone deacetylases. Defects in ISGF3 can cause resistance to IFN-.(2a) treatment.

Sequence

Synthetic peptide located within the following region: PWKHAGKQDFREDQDAAFFKAWAIFKGKYKEGDTGGPAVWKTRLRCALNK

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hou-Zao Chen et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(36), 11897-11912 (2014-09-05)
The failure of past efforts to develop effective stroke treatments is at least partially because these treatments often interfered with essential physiological functions, even though they are targeted toward pathophysiological events, such as inflammation, excitotoxicity, and oxidative stress. Thus, the
Shinichi Kadota et al.
Nucleic acids research, 42(12), 7642-7653 (2014-06-01)
Chromatin structure and its alteration play critical roles in the regulation of transcription. However, the transcriptional silencing mechanism with regard to the chromatin structure at an unstimulated state of the interferon (IFN)-stimulated gene (ISG) remains unclear. Here we investigated the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service