Skip to Content
Merck
All Photos(3)

Documents

AV09021

Sigma-Aldrich

Anti-CAV3 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Caveolin 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

17 kDa

species reactivity

bovine, human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CAV3(859)

Immunogen

Synthetic peptide directed towards the N terminal region of human CAV3

Application

Anti-CAV3 antibody produced in rabbit is suitable for western blotting at a concentration of 0.5 μg/ml.

Biochem/physiol Actions

Caveolin-3 is a membrane protein that binds to the Ca(2+)-release channel, RyR1 and facilitates caveolae formation and Ca+2 homeostasis. CAV3 regulates human ether-a-go-go-related gene (hERG) channels that controls cardiac repolarization.

Target description

CAV-3 is a meber of the caveolin family of proteins. It functions as a scaffolding protein caveolin-interacting components and is found highly expressed in muscle.

Sequence

Synthetic peptide located within the following region: MMAEEHTDLEAQIVKDIHCKEIDLVNRDPKNINEDIVKVDFEDVIAEPVG

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gareth Whiteley et al.
The Journal of biological chemistry, 287(48), 40302-40316 (2012-10-17)
Caveolin-3 facilitates both caveolae formation and a range of cell signaling pathways, including Ca(2+) homeostasis. Caveolin-3 forms a disc-shaped nonamer that binds the Ca(2+)-release channel, RyR1. Multiple caveolin-3 nonamers bind to a single RyR1 homotetramer. First three-dimensional structural insights into
Jun Guo et al.
The Journal of biological chemistry, 287(40), 33132-33141 (2012-08-11)
The human ether-a-go-go-related gene (hERG) encodes the rapidly activating delayed rectifier potassium channel (I(Kr)) which plays an important role in cardiac repolarization. A reduction or increase in hERG current can cause long or short QT syndrome, respectively, leading to fatal
Fan Deng et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 44(1), 279-292 (2017-11-14)
Hearts from diabetic subjects are susceptible to myocardial ischemia reperfusion (I/R) injury. Propofol has been shown to protect against myocardial I/R injury due to its antioxidant properties while the underlying mechanism remained incompletely understood. Thus, this study aimed to determine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service