Skip to Content
Merck
All Photos(2)

Key Documents

AV03032

Sigma-Aldrich

Anti-CCNG2 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Cyclin G2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$2,577.00

R$2,577.00


Estimated to ship onMay 21, 2025



Select a Size

Change View
100 μL
R$2,577.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$2,577.00


Estimated to ship onMay 21, 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

39 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CCNG2(901)

Related Categories

Immunogen

Synthetic peptide directed towards the N terminal region of human CCNG2

Application

Anti-CCNG2 antibody is suitable for western blotting at a concentration of 2.5 μg/ml.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

Cyclin G2 is an unconventional cyclin protein that shuttles between nucleus and cytoplasm and is linked to growth inhibition and cell differentiation. Overexpression of CyclinG2 results in p53-dependent arrest of cell cycle in G1/S phase. It enforces G2/M cell cycle arrest in collaboration with DNA damage checkpoint signaling.

Sequence

Synthetic peptide located within the following region: MKDLGAEHLAGHEGVQLLGLLNVYLEQEERFQPREKGLSLIEATPENDNT

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Angela M Svendsen et al.
The Biochemical journal, 457(1), 69-77 (2013-09-26)
The mechanisms whereby insulin analogues may cause enhanced mitogenicity through activation of either the IR (insulin receptor) or the IGF-IR (insulin-like growth factor 1 receptor) are incompletely understood. We demonstrate that in L6 myoblasts expressing only IGF-IRs as well as
Maike Zimmermann et al.
The Journal of biological chemistry, 287(27), 22838-22853 (2012-05-17)
To maintain genomic integrity DNA damage response (DDR), signaling pathways have evolved that restrict cellular replication and allow time for DNA repair. CCNG2 encodes an unconventional cyclin homolog, cyclin G2 (CycG2), linked to growth inhibition. Its expression is repressed by
Ashish Lal et al.
PLoS genetics, 7(11), e1002363-e1002363 (2011-11-22)
A simple biochemical method to isolate mRNAs pulled down with a transfected, biotinylated microRNA was used to identify direct target genes of miR-34a, a tumor suppressor gene. The method reidentified most of the known miR-34a regulated genes expressed in K562

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service