Skip to Content
Merck
All Photos(1)

Key Documents

AV02040

Sigma-Aldrich

Anti-TNFSF10 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Tumor necrosis factor (ligand) superfamily, member 10

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$3,042.00

R$3,042.00


Estimated to ship onApril 15, 2025



Select a Size

Change View
100 μL
R$3,042.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$3,042.00


Estimated to ship onApril 15, 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

33 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TNFSF10(8743)

General description

TNFSF10 (TRAIL) is a p53 target gene that regulates p53-dependent apoptosis.
Rabbit Anti-TNFSF10 antibody binds to human, canine, and pig TNFSF10.

Immunogen

Synthetic peptide directed towards the N terminal region of human TNFSF10

Application

Rabbit Anti-TNFSF10 antibody can be used for western blot assays at 1.0μg/ml.

Sequence

Synthetic peptide located within the following region: CFLKEDDSYWDPNDEESMNSPCWQVKWQLRQLVRKMILRTSEETISTVQE

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kageaki Kuribayashi et al.
Cancer biology & therapy, 7(12), 2034-2038 (2008-12-25)
We have identified TNFSF10 (TRAIL) as a p53-transcriptional target gene. There are two p53 DNA-binding sites in the human TNFSF10 promoter region, at 346 and 625 bp upstream of the transcription start site. A human p53-expressing adenovirus (Ad-p53) induced TRAIL

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service