APREST81169
PrEST Antigen HECTD4
Prestige Antigens™ Powered by Atlas Antibodies, buffered aqueous solution
Synonym(s):
C12orf51, FLJ34154, KIAA0614
About This Item
recombinant
expressed in E. coli
Assay
>80% (SDS-PAGE)
form
buffered aqueous solution
mol wt
predicted mol wt 29 kDa
purified by
immobilized metal affinity chromatography (IMAC)
concentration
≥0.5 mg/mL
immunogen sequence
KETVHIPGARCLYLRFDSRCSSQYDYDKLVIYAGPNTNSRKVAEYGGNTLGYGSRSVLGTGWPKDLVKVEGDTVTFSFEMRSGREHNTPDKAMWGFACTVR
Ensembl | human accession no.
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... HECTD4(283450)
General description
Application
Linkage
Physical form
Preparation Note
Legal Information
Disclaimer
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service