APREST72017
PrEST Antigen B4GALNT3
Prestige Antigens™ Powered by Atlas Antibodies, buffered aqueous solution
Synonym(s):
B4GalNac-T3, FLJ16224, FLJ40362
About This Item
recombinant
expressed in E. coli
Assay
>80% (SDS-PAGE)
form
buffered aqueous solution
mol wt
predicted mol wt 34 kDa
purified by
immobilized metal affinity chromatography (IMAC)
concentration
≥0.5 mg/mL
immunogen sequence
ETHNKCFYQENAYYQDRFSFQEYIKIDQPEKQGLEQPGFEENLLEESQYGEVAEETPASNNQNARMLEGRQTPASTLEQDATDYRLRSLRKLLAQPREGLLAPFSKRNSTASFPGRTSHIPVQQPEKRKQKPSPEPSQDSPHSD
Ensembl | human accession no.
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... B4GALNT3(283358)
General description
Application
Linkage
Physical form
Preparation Note
Legal Information
Disclaimer
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service