Skip to Content
Merck
All Photos(1)

Key Documents

APREST71076

Sigma-Aldrich

PrEST Antigen XRCC4

Prestige Antigens Powered by Atlas Antibodies, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

recombinant

expressed in E. coli

Assay

>80% (SDS-PAGE)

form

buffered aqueous solution

mol wt

predicted mol wt 33 kDa

purified by

immobilized metal affinity chromatography (IMAC)

concentration

≥0.5 mg/mL

immunogen sequence

KCVSAKEALETDLYKRFILVLNEKKTKIRSLHNKLLNAAQEREKDIKQEGETAICSEMTADRDPVYDESTDEESENQTDLSGLASAAVSKDDSIISSLDVTDIAPSRKRRQRMQRNLGTEPKMAPQENQLQEK

Ensembl | human accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... XRCC4(7518)

General description

Recombinant protein fragment of Human XRCC4 with N-terminal His6ABP (ABP = Albumin Binding Protein derived from Streptococcal Protein G) fusion tag. X-ray repair cross-complementing protein 4 (XRCC4) is also known as DNA repair protein XRCC4. It is mapped to human chromosome locus 5q14.2. It mediates joining of DNA double strand breaks during DNA repair.

Application

Suitable as a blocking agent using corresponding antibodies.

Biochem/physiol Actions

X-ray repair cross-complementing protein 4 (XRCC4) interacts with XRCC4-like factor (XLF) and DNA ligase IV in mediating nonhomologous end joining (NHEJ) cascade in DNA repair. A mutation in the XRCC4 is associated with adult-onset progressive encephalocardiomyopathy and ataxia. Patients with polymorphisms in XRCC4 gene are susceptible to schizophrenia. Single nucleotide polymorphisms in XRCC4 associates with high risk for cancer development. Mutations in XRCC4 leads to growth failure resulting in short stature accompanied by facial dysmorphism.(10)

Linkage

Corresponding Antibody HPA006801.

Physical form

Solution in 1 M urea-PBS, pH 7.4

Preparation Note

The protein solution should be gently mixed before use. Optimal concentrations and conditions for each application should be determined by the user.

Legal Information

Prestige Antigens is a trademark of Sigma-Aldrich Co. LLC

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An updated meta-analysis of XRCC4 polymorphisms and cancer risk based on 31 case-control studies
Shao N, et al.
Cancer Biomarkers : Section A of Disease Markers, 12(1), 37-47 (2013)
Mutations in XRCC4 cause primary microcephaly, short stature and increased genomic instability
Rosin N, et al.
Human Molecular Genetics, 24(13), 3708-3717 (2015)
Sliding sleeves of XRCC4-XLF bridge DNA and connect fragments of broken DNA
Brouwer I, et al.
Nature, 535(7613), 566-566 (2016)
XRCC4 rs6869366 polymorphism is associated with susceptibility to both nicotine dependence and/or schizophrenia
Pehlivan S, et al.
Archives of Clinical Psychiatry, 45(3), 53-56 (2018)
A nonsense mutation of human XRCC4 is associated with adult-onset progressive encephalocardiomyopathy
Bee L, et al.
EMBO Molecular Medicine, 7(7), 918-929 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service