Skip to Content
Merck
All Photos(1)

Key Documents

A9424

Sigma-Aldrich

Abietic acid

≥65% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H30O2
CAS Number:
Molecular Weight:
302.45
Beilstein:
2221451
EC Number:
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

concentration

≥65% (GC)

mp

139-142 °C (lit.)

SMILES string

CC(C)C1=CC2=CC[C@@H]3[C@](C)(CCC[C@@]3(C)C(O)=O)[C@H]2CC1

InChI

1S/C20H30O2/c1-13(2)14-6-8-16-15(12-14)7-9-17-19(16,3)10-5-11-20(17,4)18(21)22/h7,12-13,16-17H,5-6,8-11H2,1-4H3,(H,21,22)/t16-,17+,19+,20+/m0/s1

InChI key

RSWGJHLUYNHPMX-ONCXSQPRSA-N

Gene Information

human ... TNF(7124)

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alan Talevi et al.
Bioorganic & medicinal chemistry letters, 17(6), 1684-1690 (2007-01-20)
Linear discriminant analysis was performed to derive discriminant functions based on 2D descriptors and capable of classifying anticonvulsant from non-anticonvulsant compounds. Through application in virtual screening of the discriminant function which performed best in the validation steps, abietic acid was
Patrick Frank et al.
Analytical chemistry, 84(10), 4419-4428 (2012-05-02)
In 2008 the rostrum from an ancient warship was recovered from the Mediterranean near Acqualadrone, Sicily. To establish its provenance and condition, samples of black and brown rostrum wood were examined using sulfur K-edge X-ray absorption spectroscopy (XAS) and gas
Yi He et al.
Journal of pharmaceutical and biomedical analysis, 66, 345-348 (2012-04-10)
An herbal drug for the treatment of psoriasis showed severe clinical adverse reactions. The main component of adulterant was isolated from the drug and its chemical structure was elucidated as abietic acid using NMR, HR-MS, and HPLC-DAD-MS. Although abietic acid
Simon Janocha et al.
Chembiochem : a European journal of chemical biology, 14(4), 467-473 (2013-02-02)
Cytochrome P450s are very versatile enzymes with great potential for biotechnological applications because of their ability to oxidize unactivated CH bonds. CYP105A1 from Streptomyces griseolus was first described as a herbicide-inducible sulfonylurea hydroxylase, but it is also able to convert
Roberto E Di Paolo et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 9(15), 2214-2220 (2008-10-03)
The synthesis and photophysical properties of six new abietic acid based amine end-capped p-phenylenevinylene trimers (AECPV3) in their lowest excited singlet states are presented. The AECPV3 compounds show a large red-shift of both the absorption (25-30 nm) and emission (37-42

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service