Skip to Content
Merck
All Photos(1)

Key Documents

A9420

Supelco

6-Acetyl-7-deacetylforskolin

from Coleus forskohlii, ≥98%

Synonym(s):

6β-Acetoxy-1α,7β,9α-trihydroxy-8,13-epoxy-labd-14-en-11-one, Isoforskolin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H34O7
CAS Number:
Molecular Weight:
410.50
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

biological source

Coleus forskohlii

Assay

≥98%

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
veterinary

storage temp.

−20°C

SMILES string

CC(=O)O[C@@H]1[C@H](O)[C@@]2(C)O[C@](C)(CC(=O)[C@]2(O)[C@@]3(C)[C@@H](O)CCC(C)(C)C13)C=C

InChI

1S/C22H34O7/c1-8-19(5)11-14(25)22(27)20(6)13(24)9-10-18(3,4)16(20)15(28-12(2)23)17(26)21(22,7)29-19/h8,13,15-17,24,26-27H,1,9-11H2,2-7H3/t13-,15-,16?,17-,19-,20-,21+,22-/m0/s1

InChI key

CLOQVZCSBYBUPB-VDCUXWMXSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Antihypertensive with positive inotropic and adenyl cyclase activating properties

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S V Bhat et al.
Journal of medicinal chemistry, 26(4), 486-492 (1983-04-01)
Four naturally occurring analogues of forskolin were isolated. Forty-nine semisynthetic derivatives were prepared, incorporating structural alterations at the 1-, 6-, 7-, 9-, 11-, and 14/15-positions. Blood pressure lowering properties of 53 compounds were assessed in anesthetized normotensive cats and of
Bhat, S.V., et al.
Tetrahedron Letters, 1669-1669 (1977)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service