Skip to Content
Merck
All Photos(2)

Key Documents

A8675

Sigma-Aldrich

p-Amidinophenyl p-(6-amidino-2-indolyl)phenyl ether dihydrochloride

Powder

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$5,080.00

R$5,080.00


Estimated to ship onMay 08, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
R$5,080.00

About This Item

Empirical Formula (Hill Notation):
C22H19N5O · 2HCl
CAS Number:
Molecular Weight:
442.34
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

R$5,080.00


Estimated to ship onMay 08, 2025


Request a Bulk Order

Product Name

p-Amidinophenyl p-(6-amidino-2-indolyl)phenyl ether dihydrochloride,

form

powder

Quality Level

composition

Dye content, ≥85%

color

yellow-brown to brown

solubility

H2O: 10 mg/mL, clear, yellow to orange

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Cl.NC(=N)c1ccc(Oc2ccc(cc2)-c3cc4ccc(cc4[nH]3)C(N)=N)cc1

InChI

1S/C22H19N5O.ClH/c23-21(24)14-5-9-18(10-6-14)28-17-7-3-13(4-8-17)19-11-15-1-2-16(22(25)26)12-20(15)27-19;/h1-12,27H,(H3,23,24)(H3,25,26);1H

InChI key

FPSDFFVBCVBVKB-UHFFFAOYSA-N

Linkage

This material has a structure similar to Granular Blue (CAS No. 71431-29-3). It has been referred to in some literature as Granular Blue, a fluorescent neuronal tracer.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kuypers, H.G.J.M. and Huisman, A.M.
Adv. Cellular Neurobiol., 5, 307-307 (1984)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service