Skip to Content
Merck
All Photos(1)

Key Documents

A7730

Sigma-Aldrich

A-331440 dihydrochloride

≥98% (HPLC), solid

Synonym(s):

4’-[3-[(3(R)-Dimethylamino-1-pyrrolidinyl]propoxy]-[1,1-biphenyl]-4‘-carbonitrile dihydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$1,411.00
25 MG
R$4,391.00

R$1,411.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
R$1,411.00
25 MG
R$4,391.00

About This Item

Empirical Formula (Hill Notation):
C22H27N3O · 2HCl
CAS Number:
Molecular Weight:
422.39
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

R$1,411.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

solid

storage condition

desiccated

color

white to off-white

solubility

H2O: ≥10 mg/mL

originator

Abbott

storage temp.

2-8°C

SMILES string

CN([C@H]1CN(CCCOC2=CC=C(C3=CC=C(C#N)C=C3)C=C2)CC1)C.[H]Cl.[H]Cl

InChI

1S/C22H27N3O.2ClH/c1-24(2)21-12-14-25(17-21)13-3-15-26-22-10-8-20(9-11-22)19-6-4-18(16-23)5-7-19;;/h4-11,21H,3,12-15,17H2,1-2H3;2*1H/t21-;;/m1../s1

InChI key

XFPYVTSGYYMTFS-GHVWMZMZSA-N

Biochem/physiol Actions

A-331440 dihydrochloride is a non-imidazole H3 histamine receptor antagonist.
A-331440 dihydrochloride is a non-imidazole H3 histamine receptor antagonist. Presynaptic histamine H(3) receptors regulate release of histamine and other neurotransmitters, and histamine H(3) receptor antagonists enhance neurotransmitter release. A-331440 is a histamine H(3) receptor antagonist, which binds potently and selectively to both human and rat histamine H(3) receptors (K(i)=25 nM). Mice on a high-fat diet (45 kcal % lard) prior to 28-day oral b.i.d. dosing for measurement of obesity-related parameters. A-331440 administered at 0.5 mg/kg had no significant effect on weight, whereas 5 mg/kg decreased weight comparably to dexfenfluramine (10 mg/kg). A-331440 administered at 15 mg/kg reduced weight to a level comparable to mice on the low-fat diet. The two higher doses reduced body fat and the highest dose also normalized an insulin tolerance test. Both data shows that the histamine H(3) receptor antagonist, A-331440, has potential as an antiobesity agent.

Features and Benefits

This compound is featured on the Histamine Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Abbott. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offers many products related to histamine receptors for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service