Skip to Content
Merck
All Photos(1)

Key Documents

A7636

Sigma-Aldrich

Adenosine hemisulfate salt

≥98%

Synonym(s):

9-β-D-Ribofuranosyladenine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H13N5O4 · 1/2H2SO4
CAS Number:
Molecular Weight:
316.28
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98%

storage temp.

−20°C

SMILES string

OS(O)(=O)=O.Nc1ncnc2n(cnc12)[C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O.Nc4ncnc5n(cnc45)[C@@H]6O[C@H](CO)[C@@H](O)[C@H]6O

InChI

1S/2C10H13N5O4.H2O4S/c2*11-8-5-9(13-2-12-8)15(3-14-5)10-7(18)6(17)4(1-16)19-10;1-5(2,3)4/h2*2-4,6-7,10,16-18H,1H2,(H2,11,12,13);(H2,1,2,3,4)/t2*4-,6-,7-,10-;/m11./s1

InChI key

NJVOFRGMMVQTLK-JZBGLOBFSA-N

Biochem/physiol Actions

Endogenous neurotransmitter at adenosine receptors.
Endogenous neurotransmitter at adenosine receptors. Cardioprotective effects may relate to activation of A1 adenosine receptors. The antiplatelet and anti-inflammatory actions of adenosine appear to be mediated via the A2 adenosine receptor. In contrast, adenosine appears to be a pro-inflammatory mediator in asthma and chronic obstructive pulmonary disease (COPD).

Linkage

Do not confuse with adenosine 5′-monosulfate.

comparable product

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service