Skip to Content
Merck
All Photos(1)

Key Documents

A7019

Sigma-Aldrich

(±)Amethopterin hydrate

≥95%, powder

Synonym(s):

4-Amino-10-methylfolic acid, DL-4-Amino-N10-methylpteroylglutamic acid, MTX, Methylaminopterin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H22N8O5 · xH2O
CAS Number:
Molecular Weight:
454.44 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥95%

form

powder

color

yellow to yellow with an orange cast

solubility

H2O: insoluble

storage temp.

−20°C

SMILES string

O.CN(Cc1cnc2nc(N)nc(N)c2n1)c3ccc(cc3)C(=O)NC(CCC(O)=O)C(O)=O

Gene Information

Application

Amethopterin is a folic acid antagonist and potent anticancer agent. Amethopterin blocks DNA synthesis by preventing the production of tetrahydrofolate cofactors which are required to make thymidine. Amethopterin is actively transported into cells by the folate transporter where it is converted to a high molecular weight polyglutamate metabolite by folylpolyglutamate synthase, a dihydrofolate reductase inhibitor.

Biochem/physiol Actions

Amethopterin is a folic acid antagonist and potent anticancer agent. Blocks DNA synthesis by blocking the production of tetrahydrofolate covactors required for the synthesis of thymidine. Amethopterin is actively transported into cells by the folate transporter. In the cell, it is converted to a high molecular weight polyglutamate metabolite by folylpolyglutamate synthase that, in turn, binds to dihydrofolate reductase and inhibits its activity. Amethopterin-polyglutamate is degraded intracellularly by γ-glutamyl hydrolase.

Reconstitution

Add a minimal amount of 0.1 N NaOH to solubilize, then dilute in neutral buffer or saline. Diluted stock is stable for 1 week refrigerated and for 1 month frozen at −20 °C. References: Cell & Tissue Culture: Laboratory Procedures, Chapter 27, Page 27D:3.2 (A. Doyle, et al., eds, John Wiley) and Cell Biology: A Laboratory Handbook, vol. 2, page 417 (Julio E. Celis, ed., Academic Press).

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christian S Kaas et al.
Biotechnology journal, 10(7), 1081-1089 (2015-05-13)
Coagulation factor VIII (FVIII) is one of the most complex biopharmaceuticals due to the large size, poor protein stability and extensive post-translational modifications. As a consequence, efficient production of FVIII in mammalian cells poses a major challenge, with typical yields
Joshua F Baker et al.
Annals of the rheumatic diseases, 73(11), 1968-1974 (2013-08-02)
To determine if early MRI measures predict X-ray progression at 1 and 2 years in a large RA trial cohort. This study included 256 methotrexate (MTX)-naïve RA patients from a randomised placebo-controlled trial of golimumab (GO-BEFORE). MRIs of wrist and 2nd-5th
Roy Fleischmann et al.
Annals of the rheumatic diseases, 74(6), 1132-1137 (2014-08-22)
Disease Activity Score in 28 joints calculated with C-reactive protein (DAS28-CRP) is used instead of erythrocyte sedimentation rate (DAS28-ESR) to assess rheumatoid arthritis disease activity; however, values for remission and low disease activity (LDA) for DAS28-CRP have not been validated.
Axel Finckh et al.
Arthritis research & therapy, 16(5), 458-458 (2014-10-16)
Calcium pyrophosphate deposition (CPPD) may cause severe arthropathy, major joint destruction and treatment options are limited. The aim of this study was to test the therapeutic efficacy of methotrexate (MTX) in chronic or recurrent CPPD arthropathy. Patients with CPPD arthropathy
Anna-Birgitte Aga et al.
Annals of the rheumatic diseases, 74(2), 381-388 (2013-11-29)
To investigate whether baseline disease activity levels and responses in patients with rheumatoid arthritis (RA) changed during the period 2000-2010. Data were provided by the Norwegian disease-modifying antirheumatic drug (NOR-DMARD) study. Patients with inflammatory joint diseases starting new treatment with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service