Skip to Content
Merck
All Photos(1)

Documents

A6883

Sigma-Aldrich

Atropine methyl bromide

≥99% (TLC), powder

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H26NO3 · Br
CAS Number:
Molecular Weight:
384.31
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥99% (TLC)

form

powder

color

white to off-white

solubility

H2O: 50 mg/mL

SMILES string

Br.C[N]1(C)[C@@H]2CC[C@H]1C[C@@H](C2)OC(=O)C(CO)c3ccccc3

InChI

1S/C18H26NO3.BrH/c1-19(2)14-8-9-15(19)11-16(10-14)22-18(21)17(12-20)13-6-4-3-5-7-13;/h3-7,14-17,20H,8-12H2,1-2H3;1H/t14-,15+,16?,17?;

InChI key

YBFLMRSYEWDJEJ-ZNHDNBJUSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Competitive muscarinic acetylcholine receptor antagonist that does not cross the blood-brain barrier.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M M Knuepfer et al.
The American journal of physiology, 276(1 Pt 2), R103-R112 (1999-01-14)
It has been suggested that toxicity to cocaine is related to the relative rate of cocaine metabolism by cholinesterases and to activation of cholinergic receptors either directly or by reflex mechanisms. We examined these possibilities by altering cholinesterase activity and
Timothy A Yap et al.
Molecular cancer therapeutics, 10(2), 360-371 (2010-12-31)
AKT is frequently deregulated in cancer, making it an attractive anticancer drug target. CCT128930 is a novel ATP-competitive AKT inhibitor discovered using fragment- and structure-based approaches. It is a potent, advanced lead pyrrolopyrimidine compound exhibiting selectivity for AKT over PKA
H Ohnuma et al.
Journal of the autonomic nervous system, 57(1-2), 43-48 (1996-02-05)
Hepatic glucoreceptor-vagal afferent inputs to the central nervous system and pancreatic vagal efferent stimuli are important for insulin secretion. In the present study, we examined the effect of intracerebroventricular (i.c.v.) injection of atropine methyl bromide (methylatropine) on the insulin response
G J Kirouac et al.
The American journal of physiology, 273(6 Pt 2), H2549-H2557 (1998-01-22)
Experiments were done in alpha-chloralose-anesthetized, paralyzed, and artificially ventilated rats to investigate the effect of L-glutamate (Glu) stimulation of the substantia nigra (SN) and ventral tegmental area (VTA) on arterial pressure (AP) and heart rate (HR). Glu stimulation of the
Siew Kian Tai et al.
Hippocampus, 22(4), 914-925 (2011-05-05)
The vestibular system has been suggested to participate in spatial navigation, a function ascribed to the hippocampus. Vestibular stimulation during spatial navigation activates a hippocampal theta rhythm (4-10 Hz), which may enhance spatial processing and motor response. We hypothesize that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service