Skip to Content
Merck
All Photos(1)

Key Documents

A5352

Sigma-Aldrich

L-(+)-2-Amino-6-phosphonohexanoic acid

≥98% (HPLC), solid

Synonym(s):

L-AP6

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14NO5P
CAS Number:
Molecular Weight:
211.15
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

color

white

SMILES string

N[C@@H](CCCCP(O)(O)=O)C(O)=O

InChI

1S/C6H14NO5P/c7-5(6(8)9)3-1-2-4-13(10,11)12/h5H,1-4,7H2,(H,8,9)(H2,10,11,12)/t5-/m0/s1

InChI key

QIOXWRQXHFVNLV-YFKPBYRVSA-N

Biochem/physiol Actions

L-(+)-2-Amino-6-phosphonohexanoic acid (L-AP6) is an excitatory glutamate analogue that is able to depolarize L-quisqualic acid (QUIS)-sensitized CA1 rat hippocampal neurons.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L A Chase et al.
Neuroscience, 106(2), 287-301 (2001-09-22)
A brief exposure of hippocampal slices to L-quisqualic acid sensitizes CA1 pyramidal neurons 30-250-fold to depolarization by two classes of excitatory amino acid analogues: (1) those whose depolarizing effects are rapidly terminated following washout, e.g. L-2-amino-4-phosphonobutanoic acid (L-AP4) and L-2-amino-6-phosphonohexanoic
L A Chase et al.
Toxicology and applied pharmacology, 219(1), 1-9 (2007-01-20)
A brief exposure of hippocampal slices to L-quisqualic acid (QUIS) sensitizes CA1 pyramidal neurons 30- to 250-fold to depolarization by certain excitatory amino acids analogues, e.g., L-2-amino-6-phosphonohexanoic acid (L-AP6), and by the endogenous compound, L-cystine. This phenomenon has been termed
M K Schulte et al.
Brain research, 649(1-2), 203-207 (1994-06-27)
Brief exposure of rat hippocampal slices to quisqualic acid (QUIS) sensitizes neurons to depolarization by the alpha-amino-omega-phosphonate excitatory amino acid (EAA) analogues AP4, AP5 and AP6. These phosphonates interact with a novel QUIS-sensitized site. Whereas L-AP4 and D-AP5 cross-react with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service