Skip to Content
Merck
All Photos(2)

Key Documents

A5181

Sigma-Aldrich

Anisotropine methyl bromide

Synonym(s):

8-Methyl-3-(2-propylpentanoyloxy)tropinium bromide

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
R$1,157.00

R$1,157.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
R$1,157.00

About This Item

Empirical Formula (Hill Notation):
C17H32BrNO2
CAS Number:
Molecular Weight:
362.35
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

R$1,157.00


Check Cart for Availability

Request a Bulk Order

form

solid

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
veterinary

storage temp.

2-8°C

SMILES string

[Br-].CCCC(CCC)C(=O)O[C@H]1C[C@H]2CC[C@@H](C1)[N+]2(C)C

InChI

1S/C17H32NO2.BrH/c1-5-7-13(8-6-2)17(19)20-16-11-14-9-10-15(12-16)18(14,3)4;/h13-16H,5-12H2,1-4H3;1H/q+1;/p-1/t14-,15+,16+;

InChI key

QSFKGMJOKUZAJM-CNKDKAJDSA-M

Gene Information

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Dobrilla et al.
Scandinavian journal of gastroenterology. Supplement, 167, 65-67 (1989-01-01)
Forty outpatients with endoscopically confirmed duodenal ulcers were entered in a double blind trial. They were randomly allocated to octatropine-methyl-bromide and sulglycotide salt (GVP) or placebo. The results show that the combination of the two drugs is less efficacious than
[Preliminary clinical research on a new anti-ulcer drug].
E Mirelli et al.
La Clinica terapeutica, 89(4), 365-371 (1979-05-31)
Diazepam and octatropine-methyl-bromide interaction: protecting activity in experimental gastric-mucosal damage.
R Niada et al.
Pharmacological research communications, 12(4), 307-317 (1980-04-01)
[Pediatric use of a drug, anticholinergic and muscle relaxant on the gastrointestinal tract].
P C Gamalero et al.
La Pediatria medica e chirurgica : Medical and surgical pediatrics, 4(1-2), 79-81 (1982-01-01)
[Effectiveness of octatropine methylbromide (OMB) in the treatment of gastritis, duodenal ulcer and spastic colon--a double blind study].
W Tittor et al.
ZFA. Zeitschrift fur Allgemeinmedizin, 58(27), 1481-1484 (1982-09-30)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service