Skip to Content
Merck
All Photos(1)

Documents

A3601

Sigma-Aldrich

Anti-ADAM-19, N-Terminal antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-A Disintegrin And Metalloproteinase-19, Anti-Meltrin-β

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

concentration

~1 mg/mL

technique(s)

western blot: 1:1,000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ADAM19(8728)

General description

ADAM19 or Meltrin-beta belongs to the ADAM (a disintegrin and metalloprotease-like domain) family. ADAM19 is found in muscle, bone, muscle, lung, heart, brain, kidney, and several other cells. This protein shares homology with the fertilins (ADAMs 1 and 2). Studies have reported that ADAM19 is involved in myoblast fusion, similar to sperm-egg fusion aided by ADAMs 1 and 2. Furthermore, ADAM19 has been implicated in osteoblast and dendritic cell differentiation, as well as in the intracellular processing of neuregulin.
Rabbit anti-ADAM-19, N-Terminal antibody localizes human ADAM19. By immunoblotting against the reduced protein, the antibody recognizes bands at 60kDa and 50kDa from cell lysates.

Immunogen

synthetic peptide corresponding to the N-terminal of furin-activated human ADAM-19.

Application

Rabbit anti-ADAM-19, N-Terminal antibody can be used for western blot assays at a dilution of 1:1,000. It can also be used for chemiluminescent immunoassays at 1:5,000.

Physical form

Solution in 0.01 M phosphate buffered saline containing 50% glycerol and 0.05% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Kurohara et al.
Biochemical and biophysical research communications, 270(2), 522-527 (2001-02-07)
Meltrin beta (ADAM19) is a member of the metalloprotease-disintegrin family. We report here chromosomal mapping of the mouse and rat meltrin beta genes and cloning and analysis of the mouse upstream regulatory regions. The meltrin beta transcript shows a spatially
J Fritsche et al.
Blood, 96(2), 732-739 (2000-07-11)
The 1alpha,25-dihydroxyvitamin D(3) (1,25- [OH](2)VD(3)) modulates the differentiation of monocytic cell lines and monocytes (MOs) in vitro. Up to now several target genes of 1,25(OH)(2)VD(3) have been described in monocytic cell lines; however, little is known about target genes in
K Shirakabe et al.
The Journal of biological chemistry, 276(12), 9352-9358 (2000-12-26)
Meltrin beta/ADAM19 is a member of ADAMs (a disintegrin and metalloproteases), which are a family of membrane-anchored glycoproteins that play important roles in fertilization, myoblast fusion, neurogenesis, and proteolytic processing of several membrane-anchored proteins. The expression pattern of meltrin beta
D Inoue et al.
The Journal of biological chemistry, 273(7), 4180-4187 (1998-03-28)
Here we report the cloning and initial biochemical characterization of the mouse metalloprotease/disintegrin/cysteine-rich (MDC) protein meltrin beta and the analysis of the mRNA expression of four MDC genes (meltrin alpha, meltrin beta, mdc9, and mdc15) in bone cells, including osteoclasts
P Wei et al.
Biochemical and biophysical research communications, 280(3), 744-755 (2001-02-13)
The adamalysins are involved in proteolysis, adhesion, fusion, and intracellular signaling. Human ADAM19/adamalysin-19 (A disintegrin and metalloproteinase 19) was identified from primary dendritic cell cDNA libraries. It has a signal sequence, a pro-domain with a "cysteine-switch" residue, a metalloproteinase domain

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service