Skip to Content
Merck
All Photos(2)

Key Documents

A3555

Sigma-Aldrich

Arachidonic acid

from Porcine liver, BioReagent, suitable for cell culture, ≥99% (GC)

Synonym(s):

cis,cis,cis,cis-5,8,11,14-Eicosatetraenoic acid, Eicosa-5Z,8Z,11Z,14Z-tetraenoic acid, Immunocytophyte

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4(CH=CHCH2)4CH2CH2CO2H
CAS Number:
Molecular Weight:
304.47
Beilstein:
1713889
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

biological source

Porcine liver

product line

BioReagent

Assay

≥99% (GC)

form

liquid

technique(s)

cell culture | mammalian: suitable

color

clear colorless to very faintly yellow

refractive index

n20/D 1.4872 (lit.)

bp

169-171 °C/0.15 mmHg (lit.)

mp

−49 °C (lit.)

solubility

absolute ethanol: 10 mg/mL

density

0.922 g/mL at 25 °C (lit.)

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC(CCC/C=C\C/C=C\C/C=C\C/C=C\CCCCC)=O

InChI

1S/C20H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22/h6-7,9-10,12-13,15-16H,2-5,8,11,14,17-19H2,1H3,(H,21,22)/b7-6-,10-9-,13-12-,16-15-

InChI key

YZXBAPSDXZZRGB-DOFZRALJSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Arachidonic acid (AA) is an unsaturated ω6 fatty acid constituent of the phospholipids of cell membranes. Phospholipase A2 releases AA from the membrane phospholipids in response to inflammation. AA is subsequently metabolized to prostaglandins and thromboxanes by at least two cyclooxygenase (COX) isoforms, to leukotrienes and lipoxins by lipoxygenases, and to epoxyeicosatrienoic acids via cytochrome p450-catalyzed metabolism. AA and its metabolites play important roles in a variety of biological processes, including signal transduction, smooth muscle contraction, chemotaxis, cell proliferation and differentiation, and apoptosis. AA has been demonstrated to bind to the a subunit of G protein and inhibit the activity of Ras GTPase-activating proteins (GAPs). Cellular uptake of AA is energy dependent and involves protein-facilitated transport across the plasma membrane.
Arachidonic acid and its metabolites play important roles in a variety of biological processes, including signal transduction, smooth muscle contraction, chemotaxis, cell proliferation and differentiation, and apoptosis.
Arachidonic acid stimulates adhesion of MDA-MB-435 human metastatic cancer cells to extracellular matrix molecules (collagen IV and vitronectin) .

Packaging

Sealed ampule.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ninna Struck Rossen et al.
PloS one, 6(9), e25196-e25196 (2011-10-04)
Cell adhesion and migration are essential for the evolution, organization, and repair of living organisms. An example of a combination of these processes is the formation of new blood vessels (angiogenesis), which is mediated by a directed migration and adhesion
Zhuoming Liu et al.
The Journal of biological chemistry, 281(23), 16043-16051 (2006-04-15)
Dbs was identified in a cDNA-based expression screen for sequences that can cause malignant growth when expressed in murine fibroblasts. In previous studies we have shown that Dbs is a Rho-specific guanine nucleotide exchange factor that can activate RhoA and/or
Akinobu Hayashi et al.
Oncology letters, 8(4), 1513-1518 (2014-09-10)
Transforming growth factor β1 (TGFβ1) regulates a variety of cellular functions, including cell growth, apoptosis and differentiation. The aim of the current study was to investigate the alterations of phenotypic events in the long-term exposure of prostate cancer (PCa) cells
Song Xu et al.
Journal of virology, 86(23), 13025-13037 (2012-09-28)
Similar to other positive-sense, single-stranded RNA viruses, hepatitis C virus (HCV) replicates its genome in a remodeled intracellular membranous structure known as the membranous web (MW). To date, the process of MW formation remains unclear. It is generally acknowledged that
Donghui Liu et al.
American journal of physiology. Cell physiology, 301(3), C739-C748 (2011-07-08)
High-density lipoprotein (HDL) can induce cyclooxygenase-2 (COX-2) expression and prostacyclin I-2 (PGI-2) release in endothelial cells to exert multiple antiatherogenic functions. This effect has been attributed mainly to the role of sphingosine-1-phosphate (S1P) integrated in HDL. However, whether apolipoprotein A-I

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service