Skip to Content
Merck
All Photos(1)

Documents

A3021

Sigma-Aldrich

5-Aminosalicylic acid

95-98%, powder

Synonym(s):

5-AS, 5-Amino-2-hydroxybenzoic acid, Mesalamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H3-2-(OH)CO2H
CAS Number:
Molecular Weight:
153.14
Beilstein:
2090421
EC Number:
MDL number:
UNSPSC Code:
12352200

Assay

95-98%

form

powder

color

dark gray to brown

mp

275-280 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

Nc1ccc(O)c(c1)C(O)=O

InChI

1S/C7H7NO3/c8-4-1-2-6(9)5(3-4)7(10)11/h1-3,9H,8H2,(H,10,11)

InChI key

KBOPZPXVLCULAV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

5-Aminosalicylic acid is a peroxidase substrate suitable for use in ELISA procedures. This substrate produces a soluble end product that is brown in color and can be read spectrophotometrically at 450 nm. The reaction may be stopped with 3 N NaOH and read at 550 nm.

replaced by

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

No data available

Flash Point(C)

No data available

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Krupa Shukla et al.
Bioorganic & medicinal chemistry letters, 21(20), 6184-6187 (2011-09-06)
A series of sulfasalazine analogs were synthesized and tested for their ability to block cystine-glutamate antiporter system xc⁻ using L-[(14)C]cystine as a substrate. Replacement of sulfasalazine's diazo group with an alkyne group led to an equally potent inhibitor, 2-hydroxy-5-((4-(N-pyridin-2-ylsulfamoyl)phenyl)ethynyl)benzoic acid
Virginie Andrzejak et al.
Bioorganic & medicinal chemistry, 19(12), 3777-3786 (2011-05-27)
Growing evidence suggests a role for the endocannabinoid (EC) system, in intestinal inflammation and compounds inhibiting anandamide degradation offer a promising therapeutic option for the treatment of inflammatory bowel diseases. In this paper, we report the first series of carboxamides
Sarvesh Kumar et al.
European journal of medicinal chemistry, 46(11), 5498-5511 (2011-10-01)
In the present study, we report the design and synthesis of novel analogs of cinnamates, thiocinnamates and thionocinnamates and evaluated the potencies of these analogs to inhibit TNF-α induced ICAM-1 expression on human endothelial cells. By using whole cell-ELISA, our
Kin Y Tam et al.
Journal of medicinal chemistry, 53(1), 392-401 (2009-12-02)
The permeability characteristics of 33 amphoteric drugs (about 64% zwitterions at physiological pH) were studied using the parallel artificial membrane permeability assay (PAMPA) at pH 6.5. The PAMPA data were modified to include the paracellular permeability component found in cellular
Suneela S Dhaneshwar et al.
European journal of medicinal chemistry, 44(1), 131-142 (2008-05-13)
Mutual amide prodrugs of 4-aminosalicylic acid with D-phenylalanine and L-tryptophan were synthesized for targeted drug delivery to the inflamed gut tissue in inflammatory bowel disease. Stability studies in aqueous buffers (pH 1.2 and 7.4) showed that the synthesized prodrugs were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service