Skip to Content
Merck
All Photos(1)

Key Documents

A281

Sigma-Aldrich

Apigenin

solid

Synonym(s):

4′,5,7-Trihydroxyflavone, 5,7-Dihydroxy-2-(4-hydroxyphenyl)-4-benzopyrone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10O5
CAS Number:
Molecular Weight:
270.24
Beilstein:
262620
EC Number:
MDL number:
UNSPSC Code:
12171500

form

solid

color

yellow to green

mp

>300 °C (lit.)

solubility

DMSO: 27 mg/mL
H2O: insoluble

SMILES string

Oc1ccc(cc1)C2=CC(=O)c3c(O)cc(O)cc3O2

InChI

1S/C15H10O5/c16-9-3-1-8(2-4-9)13-7-12(19)15-11(18)5-10(17)6-14(15)20-13/h1-7,16-18H

InChI key

KZNIFHPLKGYRTM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

A plant flavonoid that has been found to inhibit cell proliferation by arresting the cell cycle at the G2/M phase. Inhibition of growth through cell cycle arrest and induction of apoptosis appear to be related to induction of p53. Inhibits PMA-mediated tumor promotion by inhibiting protein kinase C and the resulting suppression of oncogene expression. It has also been reported to inhibit topoisomerase I-catalyzed DNA re-ligation and enhance gap junctional intercellular communication.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gao-Jun Fan et al.
Bioorganic & medicinal chemistry letters, 17(24), 6720-6722 (2007-10-30)
The bioassay-guided purification of ether extracts of Alpinia officinarum led to the isolation of two new compounds 6-hydroxy-1,7-diphenyl-4-en-3-heptanone (1) and 6-(2-hydroxy-phenyl)-4-methoxy-2-pyrone (4) as well as three known compounds 1,7-diphenyl-4-en-3-heptanone (2), 1,7-diphenyl-5-methoxy-3-heptanone (3), and apigenin (5). Their structures were established on
Iris L K Wong et al.
Antimicrobial agents and chemotherapy, 51(3), 930-940 (2006-12-30)
Drug resistance by overexpression of ATP-binding cassette (ABC) transporters is an impediment in the treatment of leishmaniasis. Flavonoids are known to reverse multidrug resistance (MDR) in Leishmania and mammalian cancers by inhibiting ABC transporters. Here, we found that synthetic flavonoid
Heshu Lu et al.
Journal of medicinal chemistry, 48(3), 737-743 (2005-02-04)
Cyclin-dependent kinases (CDKs) play a central role in cell cycle control, apoptosis, transcription, and neuronal functions. They are important targets for the design of drugs with antimitotic or antineurodegenerative effects. CDK4 and CDK6 form a subfamily among the CDKs in
Kee Dong Yoon et al.
Journal of natural products, 70(12), 2029-2032 (2007-11-13)
Three new flavonoid glycosides ( 1- 3), 11-hydroxyhainanolidol ( 4), and a new dibenzylbutyrolactone lignan glycoside ( 5) were isolated from the aerial parts of Cephalotaxus koreana Nakai, along with 19 known flavonoids. The structures of the new compounds were
Manar M Salem et al.
Journal of natural products, 69(1), 43-49 (2006-01-31)
Bioactivity-guided fractionation of the root extract of Psorothamnus arborescens yielded the new isoflavone 5,7,3',4'-tetrahydroxy-2'-(3,3-dimethylallyl)isoflavone (1a) and the new 2-arylbenzofuran 2-(2'-hydroxy-4',5'-methylenedioxyphenyl)-6-methoxybenzofuran-3-carbaldehyde (2), together with seven known compounds, including three isoflavones, fremontin (3a), glycyrrhisoflavone (4a), and calycosin (5), two pterocarpans, maackiain (6)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service