Skip to Content
Merck
All Photos(1)

Key Documents

A1277

Sigma-Aldrich

Asp-Ala

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H12N2O5
CAS Number:
Molecular Weight:
204.18
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

storage temp.

−20°C

SMILES string

CC(NC(=O)C(N)CC(O)=O)C(O)=O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C H Görbitz
Acta chemica Scandinavica. Series B: Organic chemistry and biochemistry, 41(9), 679-685 (1987-10-01)
The crystal and molecular structures of the alpha- and beta-L-Asp isomers of L-aspartyl-L-alanine have been determined at 120 K using 1226 and 1609 reflections (I greater than 2.5 sigma I), respectively. The space group for the alpha-isomer is P2(1), with
alpha-L-Aspartyl-L-alanine (a neural dipeptide) enhances synaptic transmission.
R Lim et al.
Experientia, 37(2), 158-160 (1981-02-15)
A M Pistorius et al.
International journal of peptide and protein research, 42(6), 570-577 (1993-12-01)
In order to establish parameters to identify imide derivatives formed during in vitro aging of aspartyl-alanyl-containing proteins, a dipeptide and a tetrapeptide containing this sequence were acidified and heated in vacuo. The formation of succinimide derivatives could be confirmed by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service