Skip to Content
Merck
All Photos(1)

Documents

94152

Sigma-Aldrich

4-Amino-N-(2-azidoethyl)-1,8-naphthalimide

for click labeling

Synonym(s):

4-Amino-9-(2-azidoethyl)-1,8-naphthalimide, 4-Aminonaphthalimide-X-N3, 6-Amino-2-(2-azidoethyl)-1H-benz[de]isoquinoline-1,3(2H)-dione, ANA-Azide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H11N5O2
CAS Number:
Molecular Weight:
281.27
Beilstein:
19893166
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

solubility

methanol: 0.5 mg/mL, clear, faintly yellow to yellow

fluorescence

λex 440 nm; λem 540 nm±10 nm in methanol

suitability

conforms to structure for Infrared spectrum

storage temp.

−20°C

SMILES string

Nc1ccc2C(=O)N(CCN=[N+]=[N-])C(=O)c3cccc1c23

InChI

1S/C14H11N5O2/c15-11-5-4-10-12-8(11)2-1-3-9(12)13(20)19(14(10)21)7-6-17-18-16/h1-5H,6-7,15H2

InChI key

AJSSCUILIXXVCD-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service