Skip to Content
Merck
All Photos(1)

Key Documents

94069

Sigma-Aldrich

Polysucrose 400-fluorescein isothiocyanate conjugate

average mol wt 300,000-450,000

Synonym(s):

FITC-Polysucrose 400, Fluorescein isothiocyanate-Polysucrose 400 conjugate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
R$7,113.00

R$7,113.00


Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
1 G
R$7,113.00

About This Item

UNSPSC Code:
12352116
NACRES:
NA.32

R$7,113.00


Estimated to ship onJune 01, 2025


Request a Bulk Order

form

powder

mol wt

average mol wt 300,000-450,000

fluorescence

λex 496 nm; λem 521 nm in 0.1 M phosphate pH 7.0

Analysis Note

degree of substitution 0.003-0.015

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Explore FITC-labelled polysaccharides and their properties, including impact of molecular weight on molecule dimensions.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service