Skip to Content
Merck
All Photos(1)

Documents

93372

Sigma-Aldrich

Trizma® Buffer substance

pH 7.5

Synonym(s):

Tris Buffer substance, Tris(hydroxymethyl)aminomethane Buffer substance

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:

form

fine crystals

mol wt

average mol wt 150.6

pH

7.2±0.1 (37 °C, 0.05 M)
7.5±0.1 (25 °C, 0.05 M)
8.1±0.1 (5 °C, 0.05 M)

solubility

H2O: 0.67 g/mL, clear, colorless

SMILES string

Cl.NC(CO)(CO)CO

InChI

1S/C4H11NO3.ClH/c5-4(1-6,2-7)3-8;/h6-8H,1-3,5H2;1H

InChI key

QKNYBSVHEMOAJP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Trizma is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service